Skip to Content
Merck
All Photos(1)

Key Documents

SRP0698

Sigma-Aldrich

HRAS1 (G12V) Protein

recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$368.00

$368.00


Usually ships in 2 weeks.


Select a Size

Change View
50 μG
$368.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$368.00


Usually ships in 2 weeks.

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

mol wt

~84 kDa

concentration

0.1 μg/μL

accession no.

NM_003159

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HRAS1(6792)

Biochem/physiol Actions

Recombinant human RAS1 (2-186, G12V) was expressed in E.coli cells using an N-terminal His tag. The HRAS1(G12V) mutant is constitutively active. The RAS gene superfamily encodes a group of closely related 21,000 dalton (p21) proteins with special affinity for guanine nucleotides (GTP). RAS and several other cellular proteins with similar biochemical properties are collectively known as G-proteins and they play key roles in a wide variety of cellular activities, including cell growth, differentiation, secretion, and protein trafficking (1). There are three forms of RAS gene in cells termed H-RAS, N-RAS, and K-RAS. RAS proteins play a direct causal role in human cancer and in other diseases. Mutant H-RAS, N-RAS, and K-RAS occur in varying frequencies in different tumor types (2). Other members of the RAS superfamily may also contribute to cancer.

Sequence

2-186

Reconstitution

Formulated in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service