Skip to Content
Merck
All Photos(1)

Key Documents

SRP0514

Sigma-Aldrich

NAMPT (PBEF1)

recombinant, expressed in E. coli

Synonym(s):

NAMPT, Nicotinamide phosphoribosyltransferase protein, PBEF1, Pre-B Cell-Enhancing Factor, visfatin

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$729.00

$729.00


Estimated to ship onMarch 19, 2025



Select a Size

Change View
50 μG
$729.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$729.00


Estimated to ship onMarch 19, 2025


recombinant

expressed in E. coli

form

frozen liquid

mol wt

56 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NAMPT(10135)

General description

Human Nicotinamide phosphoribosyltransferase protein (NAMPT), also known as PBEF1 or visfatin (GenBank Accession No. NM_005746), a.a. 2-491 (end), no tag, MW=56 kDa, expressed in an E. coli cell expression system.

Physical form

Aqueous solution

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service