Skip to Content
Merck
All Photos(1)

Key Documents

SRP0409

Sigma-Aldrich

Hsp40 active human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

DNAJB1, DnaJ (Hsp40) homolog, subfamily B, member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

38.5 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... DNAJB1(3337)

General description

Human Heat Shock Protein 40 (GenBank Accession No. NM_006145), full length with C-terminal His-tag, MW = 38.5 kDa, expressed in an E. coli expression system.

Application

Useful for Elisa, western blot, inhibitor screening and selectivity profiling.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah J Gibbs et al.
PloS one, 4(2), e4595-e4595 (2009-02-27)
In response to a conditioning stress, the expression of a set of molecular chaperones called heat shock proteins is increased. In neurons, stress-induced and constitutively expressed molecular chaperones protect against damage induced by ischemia and neurodegenerative diseases, however the molecular
Li Z, Srivastava P.
Current Protocols in Immunology, Review, Appendix 1:Appendix 1T-Appendix 1:Appendix 1T (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service