Skip to Content
Merck
All Photos(2)

Key Documents

SRP0395

Sigma-Aldrich

SETD2 active human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

HYPB, SET domain containing 2, SET2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

60.9 kDa

packaging

pkg of 50 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SETD2(29072)

General description

SET domain containing 2 (SETD2) is a histone methyltransferase gene and is mapped on human chromosome 3p21.31, a region associated with growth arrest of cancer cells.[1][2]
Human SETD2/pGEX6P1 (GenBank Accession No. NM_014159), amino acids 1418-1714 with N-terminal GST-tag, MW= 60.9kDa, expressed in an Escherichia coli expression system.

Biochem/physiol Actions

SET domain containing 2 (SETD2) plays a vital role in trimethylation of the histone mark H3K36 (histone 3 lysine 36). Mutation of this tumor suppressor gene (TSG) is associated with the pathogenesis of various cancers including, clear cell renal cell carcinoma (cRCC), breast cancer, lung cancer, acute lymphoblastic leukemia(ALL), and glioma.[3] In addition, SETD2 gene is also involved in the development of Huntington′s disease (HD). SETD2 plays an important role in the transcriptional elongation by maintaining chromatin structure through association with hyperphosphorylated RNA polymerase II (POLR2A). SETD2 interacts with p53 and act as a tumor suppressor in breast cancer cells.[2] SETD2 is implicated in DNA double-strand break (DSB) exclusion and homologous recombination (HR) repair in human cells.[3] SETD2 in HeLa cells alters nucleosome organization via decreasing the recruitment of the FACT (facilitates chromatin transcription) complex to active genes.[4]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Histone methyltransferase SETD2 coordinates FACT recruitment with nucleosome dynamics during transcription.
Carvalho S
Nucleic Acids Research, 41(5), 2881-2893 (2013)
SETD2-dependent histone H3K36 trimethylation is required for homologous recombination repair and genome stability.
Pfister SX
Cell Reports, 7(6), 2006-2018 (2014)
The mRNA expression of SETD2 in human breast cancer: correlation with clinico-pathological parameters.
Al Sarakbi W
BMC Cancer, 9:290 (2009)
Histone methyltransferase gene SETD2 is a novel tumor suppressor gene in clear cell renal cell carcinoma.
Duns G
Cancer Research, 70(11), 4287-4291 (2010)
S Rega et al.
Molecular and cellular neurosciences, 18(1), 68-79 (2001-07-20)
Neurodegeneration in Huntington's disease (HD) is associated with an elongated glutamine tract in the widely expressed huntingtin protein. Although the pathogenic mechanisms are still unknown, the distinct physical properties of mutant huntingtin in the brain suggest that other factors including

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service