Skip to Content
Merck
All Photos(2)

Key Documents

SRP0327

Sigma-Aldrich

PADI-2 human

recombinant, expressed in baculovirus infected Sf9 cells, ≥60% (SDS-PAGE)

Synonym(s):

Peptidyl arginine deiminase, type II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥60% (SDS-PAGE)

form

aqueous solution

mol wt

77 kDa

packaging

pkg of 10 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PADI2(11240)

General description

Human PADI2, also known as Peptidyl arginine deiminase, type II, (GenBank Accession No. NM_007365), amino acids 1-665 (end) with C-terminal FLAG-tag, MW= 77kDa, expressed in a Baculovirus infected Sf9 cell expression system. Peptidyl arginine deiminase, protein-2 (PADI2) is mapped to human chromosome 1p36.[1] It is expressed in Sertoli cells at the time of testicular development.[2]

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling. PADI-2 human has been used in in vitro binding assays.[3]

Biochem/physiol Actions

Peptidyl arginine deiminase, protein-2 (PADI2) promotes protein citrullination and represses the multiplication of cells.[4] It helps to transform the amino acid arginine in a protein into amino acid citrulline. PADI2 participates in epigenetic mechanisms.[2] It is linked with rheumatoid arthritis.[5]

Physical form

Formulated in 40 mM Tris-HCl, pH 8.0, 110 mM NaCl, 2.2 mM KCl, 80 ng/uL Flag peptide, 3 mMDTT, and 20% Glycerol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sijun Dong et al.
The Journal of investigative dermatology, 124(5), 1026-1033 (2005-04-28)
Peptidylarginine deiminases (PAD) convert protein-bound arginine residues into citrulline residues in a Ca(2+) ion-dependent manner. Among the five isoforms (PAD1, 2, 3, 4, and 6) existing in rodents and humans, PAD2 is the most widely expressed in both species, tissues
PADI2 is significantly associated with rheumatoid arthritis
Chang X, et al.
Testing, 8(12), e81259-e81259 (2013)
Peptidyl arginine deiminase 2 (Padi2) is expressed in Sertoli cells in a specific manner and regulated by SOX9 during testicular development
Tsuji-H A, et al.
Scientific Reports, 8 (2018)
Yuichiro Watanabe et al.
Journal of human genetics, 54(7), 430-432 (2009-05-30)
Peptidylarginine deiminases (PADIs), five isoforms of which have been identified, catalyze the conversion of arginine residues to citrulline residues in proteins. Recent studies have revealed that abnormal activation of PADI2, the gene for which is expressed throughout the nervous system
Peptidyl arginine deiminase 2 (Padi2) is expressed in Sertoli cells in a specific manner and regulated by SOX9 during testicular development
Tsuji-H A, et al.
Scientific reports, 8 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service