Skip to Content
Merck
All Photos(1)

Key Documents

SRP0191

Sigma-Aldrich

Sphingosine-1-phosphate lyase1 Active human

recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)

Synonym(s):

SGPL1, SP-lyase 1, hSPL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

57.5 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

0.19 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SGPL1(8879)

General description

SGPL1 (sphingosine-1-phosphate lyase 1) is an ER (endoplasmic reticulum) enzyme which shows a wide level of tissue expression.[1] This protein is composed of putative 568 amino acids, and has a molecular weight of 63.5kDa. This enzyme is a member of the pyridoxal 5′-phosphate-dependent carbon-carbon lyases, which form a subclass of aldehyde lyases. It is a type III membrane protein, also known as type I membrane protein which lacks a cleavable N-terminal signal sequence. The single transmembrane segment is in proximity to the N-terminal which faces the ER lumen. The catalytic region faces the cytoplasm.[2]

Application

Useful for the study of biochemical function.

Biochem/physiol Actions

SGPL1 (sphingosine-1-phosphate lyase 1) enzyme is involved in the synthesis of long-chain aliphatic aldehydes which occurs during several metabolic processes, such as peroxisomal α-oxidation of 3-methyl-branched fatty acids and 2-hydroxy long-chain fatty acids, microsomal oxidation of long-chain alcohols, prenylated protein degradation in the lysosomes, microsomal degradation of phosphorylated sphingoid bases, attack of plasmalogens by myeloperoxidase (MPO)-derived hypochlorous acid, and degradation of (lyso)plasmalogens in the microsomes.[3]s This protein shows reduced expression in OSCC (oral squamous cell carcinoma), colon and prostate cancers, and this is associated with poor prognosis.[4]

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 10% glycerol, 3 mM DTT, 0.5 mM pyridoxal 5′ phosphate monohydrate.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sphingosine 1-phosphate lyase deficiency disrupts lipid homeostasis in liver.
Bektas M, et al.
The Journal of Biological Chemistry, 285(14), 10880-10889 (2010)
Aberrant expression of the S1P regulating enzymes, SPHK1 and SGPL1, contributes to a migratory phenotype in OSCC mediated through S1PR2.
Patmanathan SN, et al.
Scientific Reports, 6:25650 (2016)
Serena Mezzar et al.
Journal of lipid research, 55(3), 573-582 (2013-12-11)
Long-chain aldehydes are commonly produced in various processes, such as peroxisomal α-oxidation of long-chain 3-methyl-branched and 2-hydroxy fatty acids and microsomal breakdown of phosphorylated sphingoid bases. The enzymes involved in the aldehyde-generating steps of these processes are 2-hydroxyacyl-CoA lyase (HACL1)
Sphingosine 1-phosphate lyase, a key regulator of sphingosine 1-phosphate signaling and function.
Serra M and Saba JD
Advances in Enzyme Regulation, 50(1), 349-362 (2010)

Questions

  1. quelle est la séquence de la protéine ?

    1 answer
    1. Please see the link below to view the Uniprot profile of this protein, including sequence:
      https://www.uniprot.org/uniprotkb/O95470/entry#sequences

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service