Skip to Content
Merck
All Photos(1)

Key Documents

SRP0190

Sigma-Aldrich

RbAp48 Active human

recombinant, expressed in baculovirus infected insect cells, ≥40% (SDS-PAGE)

Synonym(s):

Chromatin assembly factor/CAF-1, NURF55, RBBP4, Retinoblastoma binding protein 4, Retinoblastoma-binding protein p48

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥40% (SDS-PAGE)

form

aqueous solution

mol wt

48 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RBBP4(5928)

General description

Human RbAp48 (NM_005610) (amino acids 2-end) with N-terminal His tag, MW = 48 kDa, expressed in baculovirus expression system.

Application

Useful for the western blot analysis.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 138 mM NaCl, 0.05% Tween-20, 100 μg/ml flag peptide and 10% glycerol.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service