Skip to Content
Merck
All Photos(1)

Key Documents

SRE0068

Sigma-Aldrich

SSC Buffer, 20X Concentrate

Synonym(s):

Saline Sodium Citrate Buffer

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$154.00

$154.00


Estimated to ship onApril 03, 2025


Request a Bulk Order

Select a Size

Change View
1 L
$154.00

About This Item

UNSPSC Code:
41105600
NACRES:
NA.25

$154.00


Estimated to ship onApril 03, 2025


Request a Bulk Order

form

liquid

Quality Level

application(s)

diagnostic assay manufacturing

storage temp.

2-8°C

Application

SSC Buffer, 20X Concentrate, is a standard reagent in Southern and Northern blotting procedures. This concentrated formula can be used straight or diluted to make SSC working solutions with concentrations as low as 0.5X. It is also for use in preparing nucleic acids for hybridization.

Physical form

This SSC Buffer, 20X Concentrate, contains 0.3 M sodium citrate in 3 M NaCl. Preparation of 1X SSC working buffer: Dilute the 20X concentrated buffer 20-fold with ultrapure water (= 18 MΩ x cm resistivity at 25 °C).

Notes: If precipitation is present in the 20X concentrated buffer, warm the bottle to 37 °C and mix until completely dissolved prior to dilution. It is recommended 1X working solutions be filtered through a 0.2 mm filter before use.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service