Skip to Content
Merck
All Photos(1)

Key Documents

SRE0033

Sigma-Aldrich

Tris Acetate-EDTA Buffer 50X concentrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$362.00

$362.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$362.00

About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

$362.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

application(s)

diagnostic assay manufacturing

storage temp.

room temp

Application

Running buffer for nucleic acid electrophoresis.

Physical form

2M TRIS acetate and 0.05M EDTA in water. Solution is 0.2 μm filtered.

Dilute with water to 1X.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service