Skip to Content
Merck
All Photos(1)

Key Documents

SML3944

Sigma-Aldrich

SJ11646

new

≥98% (HPLC)

Synonym(s):

N-(2-Chloro-6-methylphenyl)-2-((6-(4-(3-(2-(4-(2,6-dioxopiperidin-3-yl)phenoxy)acetamido)propyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)thiazole-5-carboxamide, SJ 1011646, SJ 11646, SJ-1011646, SJ-11646, SJ1011646

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H40ClN9O5S
CAS Number:
Molecular Weight:
746.28
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CC1=NC(NC2=NC=C(C(NC3=C(C=CC=C3Cl)C)=O)S2)=CC(N4CCN(CC4)CCCNC(COC5=CC=C(C=C5)C6CCC(NC6=O)=O)=O)=N1

InChI key

AJNQGWOQPRWRML-UHFFFAOYSA-N

Biochem/physiol Actions

Potent ABL and SRC family tyrosine kinases (TYKs) degrader with greater antileukemic efficacy than dasatinib in cultures and in vivo.



SJ11646 is a potent and high-affinity (LCK/ABL1/SRC Kd = 0.14/0.054/0.17 nM for SJ11646 vs. 0.09/0.054/0.19 nM for dasatinib) ABL and SRC family tyrosine kinases (TYKs) degrader composed of dasatinib linked to a E3 ligase cereblon (CRBN)-binding ligand. SJ11646 effectively degrades SRC family TYKs in KOPT-K1 cells (LCK, CSK, SRC, FYN; 100 nM for 24h) and is 55,114-times more cytotoxic (LC50 = 0.0123 pM) than dasatinib against the BCR-ABL fusion-harboring B-ALL cell line SUP-B15. When used at equal molar amount via daily i.p., SJ11646 (15 mg/kg) shows significantly greater antileukemic efficacy than dasatinib (10 mg/kg) in two T-ALL PDX models in mice in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianzhong Hu et al.
Science translational medicine, 14(659), eabo5228-eabo5228 (2022-08-25)
T cell acute lymphoblastic leukemia (T-ALL) is an aggressive hematological malignancy, and there is an unmet need for targeted therapies, especially for patients with relapsed disease. We have recently identified pre-T cell receptor and lymphocyte-specific protein tyrosine kinase (LCK) signaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service