Skip to Content
Merck
All Photos(2)

Key Documents

SML3723

Sigma-Aldrich

JNJ-26854165

≥98% (HPLC)

Synonym(s):

1-N-[2-(1H-Indol-3-yl)ethyl]-4-N-pyridin-4-ylbenzene-1,4-diamine, JNJ 1, JNJ 26854165, N1-[2-(1H-Indol-3-yl)ethyl]-N4-4-pyridinyl-1,4-benzenediamine, Serdemetan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20N4
CAS Number:
Molecular Weight:
328.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[nH]1c2c(c(c1)CCNc3ccc(cc3)Nc4ccncc4)cccc2

InChI key

CEGSUKYESLWKJP-UHFFFAOYSA-N

Biochem/physiol Actions

Inhibitor of the E3 ligase human double minute-2 (HDM2); cholesterol transport inhibitor
JNJ-26854165 (serdemetan) is an inhibitor of the E3 ligase human double minute-2 (HDM2) that inhibits cell proliferation and induces apoptosis in models of acute myeloid and lymphoid leukemias, and solid tumors. Also JNJ-26854165 inhibits cholesterol transport. JNJ-26854165 appears to stimulate cholesterol regulatory genes, but simultaneously inhibits cholesterol transport.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The novel anticancer agent JNJ-26854165 induces cell death through inhibition of cholesterol transport and degradation of ABCA1
Journal of Pharmacology and Experimental Therapeutics, 346(3), 381-392 (2013)
Zih-Yin Lai et al.
International journal of molecular sciences, 22(16) (2021-08-28)
As the most common gene mutation found in cancers, p53 mutations are detected in up to 96% of high-grade serous ovarian carcinoma (HGSOC). Meanwhile, mutant p53 overexpression is known to drive oncogenic phenotypes in cancer patients and to sustain the
Liangshun You et al.
Oncotarget, 8(5), 7777-7790 (2016-12-22)
Chronic myeloid leukemia (CML) is a clonal malignant disease caused by the expression of BCR/ABL. MDM2 (human homolog of the murine double minute-2) inhibitors such as Nutlin-3 have been shown to induce apoptosis in a p53-dependent manner in CML cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service