Skip to Content
Merck
All Photos(1)

Key Documents

SML3574

Sigma-Aldrich

5-Ph-IAA

≥97% (HPLC)

Synonym(s):

2-(5-Phenyl-1H-indol-3-yl)acetic acid, 5-Phenyl-1H-indole-3-acetic acid, 5-Phenyl-indole-3-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13NO2
CAS Number:
Molecular Weight:
251.28
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(CC1=CNC2=CC=C(C3=CC=CC=C3)C=C21)O

InChI key

ACCCWYHBDGKDKN-UHFFFAOYSA-N

Biochem/physiol Actions

5-Ph-IAA is a bumped auxin-inducible degron (AID) ligand that induces Skp1-Cul1-F-box (SCF) E3 ligase complex-mediated ubiquitinaton of Arabidopsis IAA17-derived mini-AID (mAID) degron-fused target proteins in cells expressing a hole-modified Oryza sativa TIR1 (OsTIR1) mutant (F74G or F74A). Unlike the traditional AID system, 5-Ph-IAA-based AID2 method shows no detectable leaky degradation, requires a 670-times lower ligand concentration, and achieves quicker degradation in cultures (0.2-5 μM) and in vivo (1-10 mg/kg via daily i.p. to mice xenografted with HCT116 CMV-OsTIR1(F74G) cells expressing mAID-tagged BRD4 or TOP2A).
Bumped AID2 system ligand that induces degradation of IAA17-derived mini-AID (mAID) degron-fused target proteins in cultures and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An engineered, orthogonal auxin analog/AtTIR1(F79G) pairing improves both specificity and efficacy of the auxin degradation system in Caenorhabditis elegans
Genetics, 220(2), iyab174-iyab174 (2022)
The enteric nervous system of the C. elegans pharynx is specified by the Sine oculis-like homeobox gene ceh-34
eLife, 11, e76003-e76003 (2022)
SIN-Like Pathway Kinases Regulate the End of Mitosis in the Methylotrophic Yeast Ogataea polymorpha
Cells, 11(9), 1519-1519 (2022)
The auxin-inducible degron 2 (AID2) system enables controlled protein knockdown during embryogenesis and development in Caenorhabditis elegans
Genetics, 220(2), iyab218-iyab218 (2022)
Aisha Yesbolatova et al.
Nature communications, 11(1), 5701-5701 (2020-11-13)
Protein knockdown using the auxin-inducible degron (AID) technology is useful to study protein function in living cells because it induces rapid depletion, which makes it possible to observe an immediate phenotype. However, the current AID system has two major drawbacks:

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service