Skip to Content
Merck
All Photos(1)

Key Documents

SML3524

Sigma-Aldrich

EtDO-P4

≥95% (HPLC)

Synonym(s):

D-EtDO-P4, D-threo-1-(3′,4′-Ethylenedioxy)phenyl-2-palmitoylamino-3-pyrrolidino-1-propanol, Ethylenedioxy-P4, N-[(1R,2R)-2-(2,3-Dihydro-1,4-benzodioxin-6-yl)-2-hydroxy-1-(1-pyrrolidinylmethyl)ethyl]hexadecanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H52N2O4
CAS Number:
Molecular Weight:
516.76
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

N3(CCCC3)C[C@@H](NC(=O)CCCCCCCCCCCCCCC)[C@H](O)c1cc2c(cc1)OCCO2

InChI

1S/C31H52N2O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-17-30(34)32-27(25-33-20-15-16-21-33)31(35)26-18-19-28-29(24-26)37-23-22-36-28/h18-19,24,27,31,35H,2-17,20-23,25H2,1H3,(H,32,34)/t27-,31-/m1/s1

InChI key

BBTZZVJOQCCAOR-DLFZDVPBSA-N

Biochem/physiol Actions

Cell penetrant, potent and selective nanomolar inhibitor of UDP-glucose ceramide glucosyltransferase (UGCG)
EtDO-P4, a ceramide analog, is a cell penetrant, potent and selective nanomolar inhibitor of UDP-glucose ceramide glucosyltransferase (UGCG). Inhibition of UGCG by EtDO-P4 decreases drug resistance and enhances cytotoxicity of chemotherapeutic drugs such as vincristine, daunorubicin, Lucena-1 and cisplatin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucosylceramide synthase inhibitors D-PDMP and D-EtDO-P4 decrease the GM3 ganglioside level, differ in their effects on insulin receptor autophosphorylation but increase Akt1 kinase phosphorylation in human hepatoma HepG2 cells
Acta Biochimica Polonica, 63 (2016)
Globoside promotes activation of ERK by interaction with the epidermal growth factor receptor
Biochimica et Biophysica Acta, 1820 (2012)
Eduardo J Salustiano et al.
The Journal of biological chemistry, 295(19), 6457-6471 (2020-04-02)
Multidrug resistance (MDR) in cancer arises from cross-resistance to structurally- and functionally-divergent chemotherapeutic drugs. In particular, MDR is characterized by increased expression and activity of ATP-binding cassette (ABC) superfamily transporters. Sphingolipids are substrates of ABC proteins in cell signaling, membrane

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service