Skip to Content
Merck
All Photos(1)

Key Documents

SML3463

Sigma-Aldrich

β,β-Dimethylacrylshikonin

≥98% (HPLC)

Synonym(s):

3-​methyl-​2-​butenoic acid (1R)​-​1-​(1,​4-​dihydro-​5,​8-​dihydroxy-​1,​4-​dioxo-​2-​naphthalenyl)​-​4-​methyl-​3-​penten-​1-​yl ester, Arnebin-1, Dimethylacrylshikonin, Dmask, [(1R)-1-(5,8-Dihydroxy-1,4-dioxonaphthalen-2-yl)-4-methylpent-3-enyl] 3-methylbut-2-enoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22O6
CAS Number:
Molecular Weight:
370.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, Red to very dark purple

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

O([C@H](CC=C(C)C)C1=CC(=O)c2c(c(ccc2O)O)C1=O)C(=O)C=C(C)C

InChI

1S/C21H22O6/c1-11(2)5-8-17(27-18(25)9-12(3)4)13-10-16(24)19-14(22)6-7-15(23)20(19)21(13)26/h5-7,9-10,17,22-23H,8H2,1-4H3/t17-/m1/s1

InChI key

BATBOVZTQBLKIL-QGZVFWFLSA-N

Biochem/physiol Actions

β,β-Dimethylacrylshikonin (Arnebin-1) is a main active naphthoquinone from herbal plants Lithospermum erythrorhizon (Zicao) and Arnebia euchroma that exhibits anti-neoplasm and antihyperglycemic activities. It stimulates autophagy in lung adenocarcinoma cells through activation of CaMKKβ-AMPK-mTOR pathway. β,β-Dimethylacrylshikonin facilitates wound healing through PI3K pathway. β,β-Dimethylacrylshikonin improves nonalcoholic fatty liver disease induced by a high-fat diet in rats.
Anti-neoplasm and antihyperglycemic; facilitates wound healing through PI3K pathway.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haibing Wang et al.
Biochemical and biophysical research communications, 517(3), 477-483 (2019-08-05)
β,β-Dimethylacrylshikonin (DMAS), an active ingredient of Lithospermum erythrorhizon and Arnebia euchroma, possess anti-neoplasm properties. Recently, DMAS was reported to stimulate autophagy in lung adenocarcinoma cells. However, the mechanisms by which DMAS modulates autophagy. have not yet been clearly elucidated. In
Weijia Yang et al.
The Journal of international medical research, 47(3), 1250-1263 (2019-01-27)
To examine the effects of arnebin-1 on nonalcoholic fatty liver disease (NAFLD) induced by a high-fat diet (HFD). Male Sprague-Dawley rats were fed an HFD for 10 weeks and then treated with arnebin-1 at a dose of 5, 10 or
Zhi Zeng et al.
International journal of molecular medicine, 36(3), 685-697 (2015-07-24)
Arnebin-1, a naphthoquinone derivative, plays a crucial role in the wound healing properties of Zicao (a traditional wound healing herbal medicine). It has been noted that Arnebin-1, in conjunction with vascular endothelial growth factor (VEGF), exerts a synergistic pro-angiogenic effect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service