Skip to Content
Merck
All Photos(1)

Key Documents

SML3381

Sigma-Aldrich

10-Hydroxy-trans-2-decenoic acid

≥98% (HPLC)

Synonym(s):

(E)-10-Hydroxy-2-decenoic acid, 10-HAD, NSC 87516, Royal jelly acid, Trans-10-hydroxy-2-decenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O3
CAS Number:
Molecular Weight:
186.25
MDL number:
UNSPSC Code:
12352106

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

OCCCCCCC\C=C\C(=O)O

InChI

1S/C10H18O3/c11-9-7-5-3-1-2-4-6-8-10(12)13/h6,8,11H,1-5,7,9H2,(H,12,13)/b8-6+

InChI key

QHBZHVUGQROELI-SOFGYWHQSA-N

Biochem/physiol Actions

10-Hydroxy-2-decenoic acid is an orally available royal jelly component that potently inhibits osteoclastogenesis. It binds to free fatty acid receptor 4 (FFAR4) on osteoclasts which inhibits RANKL (receptor activator of nuclear factor-κB (NF-κB) ligand), thereby attenuating the induction of nuclear factor of activated T cells (NFAT) c1. 10-Hydroxy-2-decenoic acid inhibits bone resorption in ovariectomized mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yosuke Tsuchiya et al.
The Journal of biological chemistry, 295(34), 12224-12232 (2020-07-11)
The supplementation of royal jelly (RJ) is known to provide a variety of health benefits, including anti-inflammatory and anti-obesity effects. RJ treatment also reportedly protects against bone loss, but no single factor in RJ has yet been identified as an
Mengmeng You et al.
European journal of pharmacology, 865, 172736-172736 (2019-10-16)
We previously reported that trans-10-hydroxy-2-decenoic acid (10-HDA), the exclusive lipid component of royal jelly (RJ), alleviates Lipopolysaccharide (LPS)-induced neuroinflammation both in vivo and in vitro. However, whether 10-HDA can protect against LPS-induced blood-brain barrier (BBB) damage is largely unexplored. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service