Skip to Content
Merck
All Photos(1)

Key Documents

SML3358

Sigma-Aldrich

AS602801

≥98% (HPLC)

Synonym(s):

1,3-Benzothiazol-2-yl(2-{[4-(morpholin-4-ylmethyl)benzyl]oxy}pyrimidin-4-yl)-acetonitrile, 2-(Benzo[d]thiazol-2-yl)-2-(2-(4-(morpholinomethyl)benzyloxy)pyrimidin-4-yl)acetonitrile, AS 602801, AS-602801, Bentamapimod, PGL 5001, PGL-5001, PGL5001

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H23N5O2S
CAS Number:
Molecular Weight:
457.55
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

[s]1c2c(nc1C(c3nc(ncc3)OCc4ccc(cc4)CN5CCOCC5)C#N)cccc2

InChI

1S/C25H23N5O2S/c26-15-20(24-28-22-3-1-2-4-23(22)33-24)21-9-10-27-25(29-21)32-17-19-7-5-18(6-8-19)16-30-11-13-31-14-12-30/h1-10,20H,11-14,16-17H2

InChI key

XCPPIJCBCWUBNT-UHFFFAOYSA-N

Biochem/physiol Actions

AS602801 (Bentamapimod, PGL5001) is an orally active, ATP-competitive, potent and selective c-Jun N-terminal kinase (JNK) inhibitor (JNK1/2/3 IC50 = 80/90/230 nM). AS602801 decreases MMP-3 upregulation in endometrium cultures from patients with endometriosis (15 μM alone or 5 μM in combination with 100-250 μM medroxyprogesterone acetate) and exhibits therapeutic efficacy in rodent endometriosis models in vivo (10-30 mg/kg via daily p.o. in mice and 10-60 mg/kg via twice daily p.o. in rats).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masahiro Yamamoto et al.
Anticancer research, 38(12), 6699-6706 (2018-12-07)
AS602801, a novel inhibitor of c-Jun N-terminal kinase (JNK), suppresses tumor initiation capacity and metastatic potential of cancer stem cells (CSCs). However, it remains unknown whether this inhibitor can chemosensitize CSCs. Using A2780 CSLC, a CSC line derived from ovarian
Maohua Chen et al.
Oncotarget, 7(45), 73903-73911 (2016-09-23)
Notch signaling is a highly conserved pathway that regulates cell fate decisions during embryonic development. Notch activation endangers neurons by modulating NF-κB and HIF-1α pathways, however, the role of Notch signaling in activating JNK/c-Jun following intracerebral hemorrhage (ICH) has not
Masahiro Yamamoto et al.
Anticancer research, 39(2), 609-617 (2019-02-04)
AS602801, an anti-cancer stem cell (CSC) candidate drug, sensitizes ovarian CSCs to paclitaxel and carboplatin by reducing the expression of survivin, an anti-apoptotic protein. The aim of the study was to examine the effect of AS602801 on the expression of
Shuai Zhang et al.
Journal of cellular and molecular medicine, 25(8), 4062-4072 (2021-02-21)
Previous studies showed that the chemotherapeutic effect of temozolomide (TMZ) and vincristine (VCR) against glioma might be blunted by the co-culture with astrocytes, and connexin-43 (CX43) was thought to play a vital role in the communication between glioma cells and
Stephen S Palmer et al.
Reproductive sciences (Thousand Oaks, Calif.), 23(1), 11-23 (2015-09-04)
Endometriosis is an estrogen (ER)-dependent gynecological disease caused by the growth of endometrial tissue at extrauterine sites. Current endocrine therapies address the estrogenic aspect of disease and offer some relief from pain but are associated with significant side effects. Immune

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service