Skip to Content
Merck
All Photos(1)

Key Documents

SML3315

Sigma-Aldrich

PF-07304814

≥98% (HPLC)

Synonym(s):

(S)-3-((S)-2-(4-Methoxy-1H-indole-2-carboxamido)-4-methylpentanamido)-2-oxo-4-((S)-2-oxopyrrolidin-3-yl)butyl dihydrogen phosphate, 4-Methoxy-N-[(1S )-3-methyl-1-[[[(1S )-2-oxo-1-[[(3S )-2-oxo-3-pyrrolidinyl]methyl]-3-(phosphonooxy)propyl]amino]carbonyl]butyl]-1H-indole-2-carboxamide, PF 007304814, PF 00835231 phosphate prodrug, PF 07304814, PF 835231 phosphate prodrug, PF-007304814, PF-835231 phosphate prodrug, PF007304814, PF07304814, PF835231 phosphate prodrug

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$86.30
25 MG
$349.00

$86.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$86.30
25 MG
$349.00

About This Item

Empirical Formula (Hill Notation):
C24H33N4O9P
CAS Number:
Molecular Weight:
552.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$86.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

PF-07304814 is a non-cytotoxic, aqueous soluble phosphate ester prodrug of the potent and selective corona virus inhibitor PF-00835231 (pro-/active drug SARS CoV-2 Ki = 174/0.27 nM). PF-07304814 is rapidly converted to PF-00835231, exhibiting anti-viral efficacy in cultures (PF-07304814/PF-00835231 IC50 with 2 μM P-pg efflux inhibitor CP-100356 = 3.8/0.24 μM (SARS2 Washington 1 & Vero E6-enACE2), 830/760 nM (SARS2 BetaCov GHB-03021/2020 & Vero E6-EGFP); PF-07304814/PF-00835231 IC50 with 0.5 μM CP-100356 = 58/80 nM (HCoV-229E & MRC-5)) without affecting P450 enzymes (>100 μM) or a range of transporters (>130 μM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Britton Boras et al.
bioRxiv : the preprint server for biology (2020-09-17)
COVID-19 caused by the SARS-CoV-2 virus has become a global pandemic. 3CL protease is a virally encoded protein that is essential across a broad spectrum of coronaviruses with no close human analogs. The designed phosphate prodrug PF-07304814 is metabolized to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service