Skip to Content
Merck
All Photos(1)

Key Documents

SML3284

Sigma-Aldrich

MS1943

≥98% (HPLC)

Synonym(s):

N-​[(1,​2-​Dihydro-​4,​6-​dimethyl-​2-​oxo-​3-​pyridinyl)​methyl]​-​1-​(1-​methylethyl)​-​6-​[6-​[4-​[2-​[(2-​tricyclo[3.3.1.13,​7]​dec-​1-​ylacetyl)​amino]​ethyl]​-​1-​piperazinyl]​-​3-​pyridinyl]​-1H-​indazole-​4-​carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H54N8O3
CAS Number:
Molecular Weight:
718.93
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Related Categories

Biochem/physiol Actions

MS1943 is a potent and selective degrader of EZH2 (enhancer of zeste homolog 2) that effectively reduces EZH2 levels in cells. MS1943 potently inhibits growth of multiple TNBC cells and in mice xenograft model.
Potent and selective degrader of EZH2 (enhancer of zeste homolog 2) that effectively reduces EZH2 levels in cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service