Skip to Content
Merck
All Photos(1)

Key Documents

SML3211

Sigma-Aldrich

Mogrol

≥98% (HPLC)

Synonym(s):

Mogrol, (3β,9β,10α,11α,24R)-9-Methyl-19-norlanost-5-ene-3,11,24,25-tetrol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$453.00

$453.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$453.00

About This Item

Empirical Formula (Hill Notation):
C30H52O4
CAS Number:
Molecular Weight:
476.73
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.77

$453.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O[C@H]1[C@@]2([C@H]([C@]4([C@@]([C@H](CC4)[C@@H](CC[C@@H](O)C(O)(C)C)C)(C1)C)C)CC=C3[C@H]2CC[C@@H](C3(C)C)O)C

InChI

1S/C30H52O4/c1-18(9-13-24(32)27(4,5)34)19-15-16-28(6)22-12-10-20-21(11-14-23(31)26(20,2)3)30(22,8)25(33)17-29(19,28)7/h10,18-19,21-25,31-34H,9,11-17H2,1-8H3/t18-,19-,21-,22+,23+,24-,25-,28+,29-,30+/m1/s1

InChI key

JLYBBRAAICDTIS-AYEHCKLZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Mogrol is an aglycone of mogrosides from Siraitia grosvenorii that exhibits neuroprotective and anticancer activities. It induces apoptosis in K562 leukemia cells through suppression of ERK1/2 and STAT3 pathways. Mogrol appears to ameliorate the memory impairment caused by Aβ1-42 in mice. Mogrol increases AMPK activity in hepatocytes and suppresses adipocyte differentiation in 3T3-L1 cells.
aglycone of mogrosides from Siraitia grosvenorii that exhibits neuroprotective and anticancer activities; suppresses adipocyte differentiation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoki Harada et al.
PloS one, 11(9), e0162252-e0162252 (2016-09-02)
This study investigated the effects of mogrol, an aglycone of mogrosides from Siraitia grosvenorii, on adipogenesis in 3T3-L1 preadipocytes. Mogrol, but not mogrosides, suppressed triglyceride accumulation by affecting early (days 0-2) and late (days 4-8), but not middle (days 2-4)
Gangling Chen et al.
The Journal of pharmacy and pharmacology, 71(5), 869-877 (2018-12-27)
Cognitive impairment is the main character of Alzheimer's disease (AD). This study mainly focused on whether mogrol, a tetracyclic triterpenoids compound of Siraitia grosvenorii Swingle, can ameliorate the memory impairment induced by Aβ1-42 . Memory impairment mice model was made
Can Liu et al.
American journal of cancer research, 5(4), 1308-1318 (2015-06-24)
Unlike solid tumors, the primary strategy for leukemia treatment is chemotherapy. However, leukemia chemotherapy is associated with adverse drug effects and drug resistance. Therefore, it is imperative to identify novel agents that effectively treat leukemia while minimizing adverse effects. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service