Skip to Content
Merck
All Photos(1)

Key Documents

SML3207

Sigma-Aldrich

MB05032

≥98% (HPLC)

Synonym(s):

2-Amino-5-isobutyl-4-[5-phosphono-2-furanyl]thiazole, 5-[2-Amino-5-(2-methylpropyl)-4-thiazolyl]-2-furanyl]phosphonic acid, CS-917 active drug, MB 05032, MB-05032, MB-06322 active drug, MB06322 active drug, [5-[2-Amino-5-(2-methylpropyl)-1,3-thiazol-4-yl]furan-2-yl]phosphonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$127.00
25 MG
$403.00

$127.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$127.00
25 MG
$403.00

About This Item

Empirical Formula (Hill Notation):
C11H15N2O4PS
CAS Number:
Molecular Weight:
302.29
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$127.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

[P](=O)(O)(O)c1[o]c(cc1)c2nc([s]c2CC(C)C)N

InChI

1S/C11H15N2O4PS/c1-6(2)5-8-10(13-11(12)19-8)7-3-4-9(17-7)18(14,15)16/h3-4,6H,5H2,1-2H3,(H2,12,13)(H2,14,15,16)

InChI key

XJMYIJPPDSZOPN-UHFFFAOYSA-N

Biochem/physiol Actions

AMP site-targeting, potent and selective fructose-1,6-bisphosphatase (FBPase) inhibitor that suppresses cellular gluconeogenesis (GNG).
MB05032 is an AMP site-targeting, potent and selective fructose-1,6-bisphosphatase (FBPase) inhibitor (rec hum liver FBPase IC50 = 16 nM, purified hum muscle/rat liver FBPase IC50 = 29/61 nM) with no potency toward glycogen phosphorylase or AMP-activated protein kinase. MB05032 inhibits cellular (rat/human hepotocytes IC50/glucose gluconeogenesis (GNG) substrate = 7.8 ?M/lactate & pyruvate/rat hepotocytes, 47.5 μM/lactate/human hepotocytes).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service