Skip to Content
Merck
All Photos(1)

Documents

SML3173

Sigma-Aldrich

Gatifloxacin sesquihydrate

≥98% (HPLC)

Synonym(s):

Gatifloxacin sesquihydrate, (±)-1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid sesquihydrate, 1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid sesquihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22FN3O4 · 1.5H2O
CAS Number:
Molecular Weight:
402.42
Beilstein:
14390028
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

O=C(O)C1=CN(C2CC2)C3=C(OC)C(N4CCNC(C)C4)=C(F)C=C3C1=O.O=C(O)C5=CN(C6CC6)C7=C(OC)C(N8CCNC(C)C8)=C(F)C=C7C5=O.O.O.O

InChI

1S/2C19H22FN3O4.3H2O/c2*1-10-8-22(6-5-21-10)16-14(20)7-12-15(18(16)27-2)23(11-3-4-11)9-13(17(12)24)19(25)26;;;/h2*7,9-11,21H,3-6,8H2,1-2H3,(H,25,26);3*1H2

InChI key

RMJMZKDEVNTXHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Gatifloxacin sesquihydrateis an oral fluoroquinolone broad spectrum antibiotic, inhibitor of bacterial enzymes DNA gyrase and topoisomerase IV, with improved gram-positive and anaerobe coverage. Gatifloxacin is reported to be useful to treat ocular infection and does not appear to cause phototoxic effects.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service