Skip to Content
Merck
All Photos(1)

Documents

SML3108

Sigma-Aldrich

Voclosporin

≥95% (HPLC)

Synonym(s):

6-[(2S,3R,4R,6E)-3-Hydroxy-4-methyl-2-(methylamino)-6,8-nonadienoic acid]cyclosporin A, Cyclosporin A, 6-[(2S,3R,4R,6E)-3-hydroxy-4-methyl-2-(methylamino)-6,8-nonadienoic acid]-, ISATX247, LX 214, R 1524

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H111N11O12
CAS Number:
Molecular Weight:
1214.62
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

film or powder

color

white to off-white

storage temp.

−20°C

InChI

1S/C63H111N11O12/c1-25-27-28-29-41(15)53(76)52-57(80)66-44(26-2)59(82)68(18)34-49(75)69(19)45(30-35(3)4)56(79)67-50(39(11)12)62(85)70(20)46(31-36(5)6)55(78)64-42(16)54(77)65-43(17)58(81)71(21)47(32-37(7)8)60(83)72(22)48(33-38(9)10)61(84)73(23)51(40(13)14)63(86)74(52)24/h25,27-28,35-48,50-53,76H,1,26,29-34H2,2-24H3,(H,64,78)(H,65,77)(H,66,80)(H,67,79)/b28-27+/t41-,42+,43-,44+,45+,46+,47+,48+,50+,51+,52+,53-/m1/s1

InChI key

BICRTLVBTLFLRD-PTWUADNWSA-N

Biochem/physiol Actions

Voclosporin is a semi-synthetic calcineurin (CN) inhibitor that exhibits potent immunosuppressive activity.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service