Skip to Content
Merck
All Photos(1)

Key Documents

SML3094

Sigma-Aldrich

SGC-SMARCA-BRDVIII

≥98% (HPLC)

Synonym(s):

1,1-Dimethylethyl 4-[3-amino-6-(2-hydroxyphenyl)-4-pyridazinyl]-1-piperazinecarboxylate, tert-Butyl 4-[3-amino-6-(2-hydroxyphenyl)pyridazin-4-yl]piperazine-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$155.00
25 MG
$494.00

$155.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$155.00
25 MG
$494.00

About This Item

Empirical Formula (Hill Notation):
C19H25N5O3
CAS Number:
Molecular Weight:
371.43
UNSPSC Code:
12352200
NACRES:
NA.77

$155.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N1CCN(CC1)C2=C(N)N=NC(C3=CC=CC=C3O)=C2)OC(C)(C)C

InChI

1S/C19H25N5O3/c1-19(2,3)27-18(26)24-10-8-23(9-11-24)15-12-14(21-22-17(15)20)13-6-4-5-7-16(13)25/h4-7,12,25H,8-11H2,1-3H3,(H2,20,22)

InChI key

AQTNUGRRZDRZIA-UHFFFAOYSA-N

Biochem/physiol Actions

Potent, SMARCA2/4 and PB1(5) BRDs-selective SWI/SNF bromodomain inhibitor that blocks adipogenesis with a >10-fold potency than PFI-3.
SGC-SMARCA-BRDVIII is a potent, SMARCA2/4 and PB1(bromo 5)-selective SWI/SNF bromodomain (BRD) inhibitor that effectively blocks adipocyte differentiation in 3T3-L1 murine fibroblast cultures (1 μM) by preventing the expression of key adipogenesis genes, including PPARγ, C/EBPα, and FABP4, with a >10-fold higher potency than PFI-3.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marek Wanior et al.
Journal of medicinal chemistry, 63(23), 14680-14699 (2020-11-21)
Accessibility of the human genome is modulated by the ATP-driven SWI/SNF chromatin remodeling multiprotein complexes BAF (BRG1/BRM-associated factor) and PBAF (polybromo-associated BAF factor), which involves reading of acetylated histone tails by the bromodomain-containing proteins SMARCA2 (BRM), SMARCA4 (BRG1), and polybromo-1.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service