Skip to Content
Merck
All Photos(1)

Key Documents

SML3078

Sigma-Aldrich

JT003 trifluoroacetate

≥95% (HPLC)

Synonym(s):

P-Nva-LYYFA trifluoroacetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$178.00

$178.00


Estimated to ship onMay 19, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$178.00

About This Item

Empirical Formula (Hill Notation):
C46H62N8O9 · xC2HF3O2
Molecular Weight:
871.03 (free base basis)
UNSPSC Code:
51111800
NACRES:
NA.77

$178.00


Estimated to ship onMay 19, 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

film or powder

color

white to off-white

storage temp.

−20°C

Biochem/physiol Actions

JT003 is potent dual agonist of adiponectin receptors AdipoR1/AdipoR2 that potently improves insulin resistance in high fat diet induced nonalcoholic steatohepatitis (NASH) mice. JT003 inhibits hepatic stellate cells (HSCs) activation in CCl4 induced liver fibrosis.
potent dual agonist of adiponectin receptors AdipoR1/AdipoR2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michishige Terasaki et al.
International journal of molecular sciences, 21(13) (2020-07-11)
Dipeptidyl peptidase-4 (DPP-4) inhibitors have been reported to play a protective role against atherosclerosis in both animal models and patients with type 2 diabetes (T2D). However, since T2D is associated with dyslipidemia, hypertension and insulin resistance, part of which are
Hongjiao Xu et al.
Nature communications, 11(1), 5807-5807 (2020-11-18)
Chronic nonalcoholic steatohepatitis (NASH) is a metabolic disorder that often leads to liver fibrosis, a condition with limited therapy options. Adiponectin is an adipocytokine that regulates glucose and lipid metabolism via binding to its receptors AdipoR1 and AdipoR2, and AdipoRs

Questions

  1. Good afternoon, I would like to know the solubility of compound JT003 for in vivo experiments on mice.  Thank you. Maria

    1 answer
    1. This product is not tested for in vivo or in vitro use. Several publication indicate that this compound has been used successfully in mouse models. Please see the links below to review publications that may be of interest.
      https://pubmed.ncbi.nlm.nih.gov/36873175/
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC7669869/

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service