Skip to Content
Merck
All Photos(1)

Key Documents

SML3061

Sigma-Aldrich

BSJ-04-122

>97% (HPLC)

Synonym(s):

N-(2-((5-chloro-7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)phenyl)acrylamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$127.00
25 MG
$510.00

$127.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$127.00
25 MG
$510.00

About This Item

Empirical Formula (Hill Notation):
C15H12ClN5O
CAS Number:
Molecular Weight:
313.74
UNSPSC Code:
12352200
NACRES:
NA.77

$127.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Quality Level

Assay

>97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

BSJ-04-122 is a cell penetrant, potent and selective covalent inhibitor of MKK4 and MKK7 (MAP2K4/7) that binds to Cys247 of MKK4. It potently inhibits JNK phosphorylation. BSJ-04-122 exhibits antiproliferative effects against triple-negative breast cancer cells. Also, BSJ-04-122 enhances antiproliferative activity of JNK-IN-8.
cell penetrant, potent and selective covalent inhibitor of MKK4 and MKK7 (MAP2K4/7)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service