Skip to Content
Merck
All Photos(1)

Key Documents

SML2987

Sigma-Aldrich

Cefcapene Pivoxil Hydrochloride Hydrate

≥98% (HPLC)

Synonym(s):

Cefcapene Pivoxil Hydrochloride Hydrate, [6R-[6α,7β(Z)]]-3-[[(Aminocarbonyl)oxy]methyl]-7-[[2-(2-amino-4-thiazolyl)-1-oxo-2-pentenyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid (2,2-dimethyl-1-oxopropoxy)methyl ester monohydrochloride monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$101.00
50 MG
$405.00

$101.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$101.00
50 MG
$405.00

About This Item

Empirical Formula (Hill Notation):
C23H29N5O8S2 · xHCl · yH2O
Molecular Weight:
567.64 (anhydrous free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

$101.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

Biochem/physiol Actions

Cefcapene Pivoxil Hydrochloride Hydrate is an orally available prodrug of third-generation cephalosporin cefcapene that exhibits potent and wide spectrum antibacterial activity against Gram-positive and Gram-negative bacteria, including Staphylococcus aureus.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin He et al.
Antimicrobial agents and chemotherapy, 48(7), 2604-2609 (2004-06-25)
The aim of this study was to elucidate the absorption mechanism in Caco-2 and rat intestine models in order to improve the accuracy of prediction of oral absorption of ester prodrugs. Pivampicillin and cefcapene pivoxil hydrochloride (CFPN-PI), ester-type oral antibiotics
Przemysław Zalewski et al.
Chromatographia, 76(7-8), 387-391 (2013-04-05)
The stability-indicating LC assay method was developed and validated for quantitative determination of cefcapene pivoxil in the presence of degradation products formed during forced degradation studies. An isocratic RP-HPLC method was developed with a Lichrospher RP-18 (250 mm × 4.6 mm, 5 μm) column and
Masamoto Murakami et al.
Dermatology (Basel, Switzerland), 231(4), 304-311 (2015-10-07)
Pustulosis palmaris et plantaris or palmoplantar pustulosis (PPP) is a refractory pustular eruption of the palms and soles with unknown etiology. In addition to skin lesions, PPP patients may present with severe joint pain and pustulotic arthro-osteitis (PAO), especially of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service