Skip to Content
Merck
All Photos(1)

Key Documents

SML2985

Sigma-Aldrich

AM-0902

≥98% (HPLC)

Synonym(s):

1-((3-(4-Chlorophenethyl)-1,2,4-oxadiazol-5-yl)methyl)-7-methyl-1H-purin-6(7H)-one, 1-[[3-[2-(4-Chlorophenyl)ethyl]-1,2,4-oxadiazol-5-yl]methyl]-1,7-dihydro-7-methyl-6H-Purin-6-one, AM 0902, AMG0902

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$117.00
25 MG
$481.00

$117.00


Estimated to ship onMay 20, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$117.00
25 MG
$481.00

About This Item

Empirical Formula (Hill Notation):
C17H15ClN6O2
CAS Number:
Molecular Weight:
370.79
UNSPSC Code:
51111800
NACRES:
NA.77

$117.00


Estimated to ship onMay 20, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)CCc2n[o]c(n2)C[n]3[c](c4[n](cnc4nc3)C)=O

InChI

1S/C17H15ClN6O2/c1-23-9-19-16-15(23)17(25)24(10-20-16)8-14-21-13(22-26-14)7-4-11-2-5-12(18)6-3-11/h2-3,5-6,9-10H,4,7-8H2,1H3

InChI key

AWJBWNUUODWOKQ-UHFFFAOYSA-N

Biochem/physiol Actions

AM-0902 (AMG0902) is a potent and selective transient receptor potential A1 (TRPA1) antagonist (rat/human TRPA1 IC50 = 71/131 nM against AITC-induced 45Ca2+ influx in respective CHO transfectants with [AITC] = EC90 = 3 μM/hTRPA1 or 35 μM/rTRPA1; human/rat IC50 = 24/20 nM by FLIPR) with little potency against human TRPV1/4, human CYP3A4/2D6, rat TRPV1/3, rat TRPM8, or mouse NaV1.7. AM-0902 effectively reduces AITC-induced flinching in a rat pain model (by 65% and 85%, respectively, with 10 or 30 mg/kg AM-0902 p.o. 1 hr prior to AITC injection) with good pharmacokinetic properties, oral availability and brain exposure (F = 60%, B/P = 0.2; 30 mg/kg p.o. in rats) in vivo.
Brain-penetrant, orally active, potent and selective transient receptor potential A1 (TRPA1) antagonist with efficacy against AITC-induced flinching in rats in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gihan S Gunaratne et al.
PLoS neglected tropical diseases, 12(4), e0006420-e0006420 (2018-04-19)
The anthelmintic praziquantel (±PZQ) serves as a highly effective antischistosomal therapy. ±PZQ causes a rapid paralysis of adult schistosome worms and deleterious effects on the worm tegument. In addition to these activities against the parasite, ±PZQ also modulates host vascular
Sonya G Lehto et al.
Molecular pain, 12 (2016-12-03)
The transient receptor potential ankyrin 1 (TRPA1) channel has been implicated in pathophysiological processes that include asthma, cough, and inflammatory pain. Agonists of TRPA1 such as mustard oil and its key component allyl isothiocyanate (AITC) cause pain and neurogenic inflammation
Laurie B Schenkel et al.
Journal of medicinal chemistry, 59(6), 2794-2809 (2016-03-05)
There has been significant interest in developing a transient receptor potential A1 (TRPA1) antagonist for the treatment of pain due to a wealth of data implicating its role in pain pathways. Despite this, identification of a potent small molecule tool
Nobuaki Takahashi et al.
Cancer cell, 33(6), 985-1003 (2018-05-29)
Cancer cell survival is dependent on oxidative-stress defenses against reactive oxygen species (ROS) that accumulate during tumorigenesis. Here, we show a non-canonical oxidative-stress defense mechanism through TRPA1, a neuronal redox-sensing Ca2+-influx channel. In TRPA1-enriched breast and lung cancer spheroids, TRPA1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service