Skip to Content
Merck
All Photos(1)

Key Documents

SML2976

Sigma-Aldrich

WR99210 hydrochloride

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$205.00
25 MG
$659.00

$205.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$205.00
25 MG
$659.00

About This Item

Empirical Formula (Hill Notation):
C14H18Cl3N5O2 · HCl
CAS Number:
Molecular Weight:
431.14
UNSPSC Code:
51111800
NACRES:
NA.77

$205.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[Cl-].Clc1c(cc(c(c1)Cl)OCCCON2C(N=C(N=C2N)N)(C)C)Cl.[H+]

InChI

1S/C14H18Cl3N5O2.ClH/c1-14(2)21-12(18)20-13(19)22(14)24-5-3-4-23-11-7-9(16)8(15)6-10(11)17;/h6-7H,3-5H2,1-2H3,(H4,18,19,20,21);1H

InChI key

VOTSDCGUYAGUNS-UHFFFAOYSA-N

Biochem/physiol Actions

Potent P. falciparum dihydrofolate reductase (pfDHFR) inhibitor.
WR99210 is a potent inhibitor of Plasmodium falciparum dihydrofolate reductase (pfDHFR), which is a major malarial drug target. It has subnanomolar potency for the wild type, double mutant and quadruple mutant dihydrofolate reductases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hekmat-Nejad et al.
Experimental parasitology, 87(3), 222-228 (1997-11-26)
With emerging drug resistance in Plasmodium falciparum, novel antifolates effective against pyrimethamine-resistant and cycloguanil-resistant dihydrofolate reductase (DHFR) are in demand. Based on structural similarity to cycloguanil, it has been proposed that WR99210, and its metabolic precursor PS-15, exerts selective antimalarial
Yvette S Levray et al.
Molecular and biochemical parasitology, 238, 111292-111292 (2020-06-09)
Defining protein-protein interactions is fundamental to the understanding of gene function. Protein-fragment complementation assays have been used for the analysis of protein-protein interactions in various organisms. The split-dihydrofolate reductase (DHFR) protein-fragment complementation assay utilises two complementary fragments of the enzyme
Jan D Warncke et al.
Cellular microbiology, 22(2), e13123-e13123 (2019-10-28)
A hallmark of the biology of Plasmodium falciparum blood stage parasites is their extensive host cell remodelling, facilitated by parasite proteins that are exported into the erythrocyte. Although this area has received extensive attention, only a few exported parasite proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service