Skip to Content
Merck
All Photos(1)

Key Documents

SML2972

Sigma-Aldrich

Cyclopentenyl uracil

≥98% (HPLC)

Synonym(s):

1-[(1R,4R,5S)-4,5-Dihydroxy-3-(hydroxymethyl)-2-cyclopenten-1-yl]-2,4(1H,3H)-pyrimidinedione, Cyclopentenyluracil, Cyclopentenyluridine, NSC 375574

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$135.00
25 MG
$541.00

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$135.00
25 MG
$541.00

About This Item

Empirical Formula (Hill Notation):
C10H12N2O5
CAS Number:
Molecular Weight:
240.21
UNSPSC Code:
51111800
NACRES:
NA.77

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[nH]1[c]([n](cc[c]1=O)[C@H]2[C@@H]([C@@H](C(=C2)CO)O)O)=O

InChI

1S/C10H12N2O5/c13-4-5-3-6(9(16)8(5)15)12-2-1-7(14)11-10(12)17/h1-3,6,8-9,13,15-16H,4H2,(H,11,14,17)/t6-,8-,9+/m1/s1

InChI key

FCWVOJUKQSHZIV-VDAHYXPESA-N

Biochem/physiol Actions

Cyclopentenyl uracil is a non-cytotoxic inhibitor of uridine kinase that act as a potent inhibitor of pyrimidine salvage in the intact mouse. Combination of Cyclopentenyl uracil with GSK983 significantly reduces viral infection of dengue serotype 2 (DENV-2) strain 16681 in cultures of the A549 lung carcinoma cell line. Cyclopentenyl uracil in combination with GSK983 enhances therapeutic index of viral RdRp inhibitor R1479.
non-cytotoxic inhibitor of uridine kinase that act as a potent inhibitor of pyrimidine salvage

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Cysyk et al.
Biochemical pharmacology, 49(2), 203-207 (1995-01-18)
Cyclopentenyl uracil, a non-cytotoxic inhibitor of uridine kinase, was found to effectively block the salvage of circulating uridine by host and tumor tissues in the intact mouse. Dose-response characteristics of the inhibition were determined. Large doses (1 g/kg) of cyclopentenyl
Qi Liu et al.
Cell chemical biology, 27(6), 668-677 (2020-05-23)
Genome-wide analysis of the mode of action of GSK983, a potent antiviral agent, led to the identification of dihydroorotate dehydrogenase as its target along with the discovery that genetic knockdown of pyrimidine salvage sensitized cells to GSK983. Because GSK983 is an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service