Skip to Content
Merck
All Photos(1)

Key Documents

SML2952

Sigma-Aldrich

Tasisulam

≥98% (HPLC)

Synonym(s):

LY 573636, LY-573636, LY573636, N-(2,4-Dichlorobenzoyl)-5-bromothiophene-2-sulfonamide, N-(5-Bromothiophen-2-ylsulfonyl)-2,4-dichlorobenzamide, N-[(5-Bromo-2-thienyl)sulfonyl]-2,4-dichlorobenzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$117.00
25 MG
$327.00

$117.00


Estimated to ship onMay 13, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$117.00
25 MG
$327.00

About This Item

Empirical Formula (Hill Notation):
C11H6BrCl2NO3S2
CAS Number:
Molecular Weight:
415.11
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$117.00


Estimated to ship onMay 13, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(NC(=O)c2c(cc(cc2)Cl)Cl)c1[s]c(cc1)Br

InChI

1S/C11H6BrCl2NO3S2/c12-9-3-4-10(19-9)20(17,18)15-11(16)7-2-1-6(13)5-8(7)14/h1-5H,(H,15,16)

InChI key

WWONFUQGBVOKOF-UHFFFAOYSA-N

Biochem/physiol Actions

RNA binding motif protein 39 (RBM39; CAPERα, HCC1) degradation-inducing SPLAM (SPLicing inhibitor sulfonAMide).
Tasisulam (LY573636) is a SPLAM (SPLicing inhibitor sulfonAMide) that bridges RNA binding motif protein 39 (RBM39; CAPERα, HCC1) and DCAF15 for interaction, thereby promoting (CUL4-DDB1-DDA1-DCAF15) E3 ubiquitin ligase complex-mediated RBM39 ubiquitination and subsequent proteasomal degradation. Tasisulam exerts selective toxicity in parental HCT116 cancer cultures (IC50 = 6.5 μM), but not those harboring non-CUL4-DCAF15-associating RBM39 G268V mutant even at a high concentration of 50 μM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Han et al.
Science (New York, N.Y.), 356(6336) (2017-03-18)
Indisulam is an aryl sulfonamide drug with selective anticancer activity. Its mechanism of action and the basis for its selectivity have so far been unknown. Here we show that indisulam promotes the recruitment of RBM39 (RNA binding motif protein 39)
Timothy Meier et al.
Molecular cancer therapeutics, 10(11), 2168-2178 (2011-09-10)
LY573636-sodium (tasisulam) is a small molecule antitumor agent with a novel mechanism of action currently being investigated in a variety of human cancers. In vitro, tasisulam induced apoptosis via the intrinsic pathway, resulting in cytochrome c release and caspase-dependent cell
Talin Haritunians et al.
Oncology reports, 20(5), 1237-1242 (2008-10-25)
LY573636-sodium is a promising anti-tumor agent, which causes growth arrest and apoptosis of a variety of human solid tumors in vitro and in vivo. Moreover, studies have shown that the compound is selectively toxic towards tumor cells over their normal
Everett J Perkins et al.
European journal of drug metabolism and pharmacokinetics, 43(3), 355-367 (2017-11-10)
Cytochrome P450 2C9 (CYP2C9) is involved in the biotransformation of many commonly used drugs, and significant drug interactions have been reported for CYP2C9 substrates. Previously published physiologically based pharmacokinetic (PBPK) models of tolbutamide are based on an assumption that its metabolic
Tyler B Faust et al.
Nature chemical biology, 16(1), 7-14 (2019-11-07)
The investigational drugs E7820, indisulam and tasisulam (aryl-sulfonamides) promote the degradation of the splicing factor RBM39 in a proteasome-dependent mechanism. While the activity critically depends on the cullin RING ligase substrate receptor DCAF15, the molecular details remain elusive. Here we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service