Skip to Content
Merck
All Photos(1)

Key Documents

SML2951

Sigma-Aldrich

SGC-CK2-1

≥98% (HPLC)

Synonym(s):

N-(5-(3-Cyano-7-(cyclopropylamino)pyrazolo[1,5-a]pyrimidin-5-ylamino)-2-methylphenyl)propionamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$210.00
25 MG
$573.00

$210.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$210.00
25 MG
$573.00

About This Item

Empirical Formula (Hill Notation):
C20H21N7O
CAS Number:
Molecular Weight:
375.43
UNSPSC Code:
12352200
NACRES:
NA.77

$210.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[n]21ncc(c2nc(cc1NC4CC4)Nc3cc(c(cc3)C)NC(=O)CC)C#N

InChI key

YKDZIFFKQUNVHH-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and highly selective casein 2 (CK2, CKII) inhibitor with potent cellular activity, but no antiproliferation efficacy in HCT116 or U-87 MG cancer cultures.
SGC-CK2-1 is a potent and highly selective casein 2 (CK2, CKII) inhibitor (CK2α/CK2α′ IC50 = 4.2/2.3 nM by cell-free enzyme assay; DYRK2 IC50 = 440 nM, IC50≥3.4 μM against 465 other kinases). Unlike less specific CK2 inhibitors, SGC-CK2-1 shows no antiproliferation activity in HCT116 or U-87 MG cancer cultures (up to 10 μM) despite its potent cellular activity (CK2α/CK2α′ IC50 = 36/16 nM by cellular NanoBRET assay).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service