Skip to Content
Merck
All Photos(1)

Key Documents

SML2900

Sigma-Aldrich

AZD5582

≥98% (HPLC)

Synonym(s):

3,3′-[2,4-Hexadiyne-1,6-diylbis[oxy[(1S,2R)-2,3-dihydro-1H-indene-2,1-diyl]]]bis[N-methyl-L-alanyl-(2S)-2-cyclohexylglycyl-L-prolinamide], AZD 5582, AZD-5582

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$149.00
25 MG
$603.00

$149.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$149.00
25 MG
$603.00

About This Item

Empirical Formula (Hill Notation):
C58H78N8O8
CAS Number:
Molecular Weight:
1015.29
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$149.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

−20°C

SMILES string

CN[C@@H](C)C(N[C@@H](C1CCCCC1)C(N2[C@H](C(N[C@H]3C4=C(C=CC=C4)C[C@H]3OCC#CC#CCO[C@@H]5CC6=C(C=CC=C6)[C@@H]5NC([C@@H]7CCCN7C([C@H](C8CCCCC8)NC([C@H](C)NC)=O)=O)=O)=O)CCC2)=O)=O

InChI

1S/C58H78N8O8/c1-37(59-3)53(67)61-49(39-21-9-7-10-22-39)57(71)65-31-19-29-45(65)55(69)63-51-43-27-15-13-25-41(43)35-47(51)73-33-17-5-6-18-34-74-48-36-42-26-14-16-28-44(42)52(48)64-56(70)46-30-20-32-66(46)58(72)50(40-23-11-8-12-24-40)62-54(68)38(2)60-4/h13

InChI key

WLMCRYCCYXHPQF-ZVMUOSSASA-N

Biochem/physiol Actions

AZD5582 is a divalent AVPI motif-based Smac (DIABLO) mimetic (SMC) that acts a potent apoptosis protein repeat (BIR) domain-targeting antagonist against inhibitor of apoptosis proteins (IAPs) cIAP1/2 (BIR3 IC50 = 15/21 nM against 2.5 nM AbuRPFK-5FAM) and XIAP (BIR2/3 IC50 = 21/15 nM). AZD5582 abolishes cellular XIAP-caspase-9 interaction (1 μM, 4 hrs), induces cIAP1/2 degradation (EC50 = 0.1 nM, 1 hr) and apoptosis (GI50 <60 pM, 48 hrs) in MDA-MB-231 breast cancer cultures. AZD5582 causes substantial tumor regression by inducing cIAP1 degradation and apoptosis in tumor cells in MDA-MB-231 xenograft-bearing mice in vivo (3.0 mg/kg/wk i.v.).
Divalent Smac mimetic compound (SMC) as a potent BIR domain-targeting antagonist against inhibitor of apoptosis proteins (IAPs) cIAP1/2 & XIAP in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward J Hennessy et al.
Journal of medicinal chemistry, 56(24), 9897-9919 (2013-12-11)
A series of dimeric compounds based on the AVPI motif of Smac were designed and prepared as antagonists of the inhibitor of apoptosis proteins (IAPs). Optimization of cellular potency, physical properties, and pharmacokinetic parameters led to the identification of compound
Jai-Hee Moon et al.
Oncotarget, 6(29), 26895-26908 (2015-09-01)
Inhibitor of apoptosis proteins (IAPs) plays an important role in controlling cancer cell survival. IAPs have therefore attracted considerable attention as potential targets in anticancer therapy. In this study, we investigated the anti-tumor effect of AZD5582, a novel small-molecule IAP
Shawn T Beug et al.
Nature communications, 8 (2017-02-16)
Small-molecule inhibitor of apoptosis (IAP) antagonists, called Smac mimetic compounds (SMCs), sensitize tumours to TNF-α-induced killing while simultaneously blocking TNF-α growth-promoting activities. SMCs also regulate several immunomodulatory properties within immune cells. We report that SMCs synergize with innate immune stimulants
Lorenz Kadletz et al.
Acta oto-laryngologica, 137(3), 320-325 (2016-10-16)
On the one hand, AZD5582, an inhibitor of inhibitor of apoptosis family proteins (IAP), leads to cellular growth arrest and induction of apoptosis in head and neck squamous cell carcinoma (HNSCC) cell lines. On the other hand, it is a
Christopher S Garris et al.
Immunity, 49(6), 1148-1161 (2018-12-16)
Anti-PD-1 immune checkpoint blockers can induce sustained clinical responses in cancer but how they function in vivo remains incompletely understood. Here, we combined intravital real-time imaging with single-cell RNA sequencing analysis and mouse models to uncover anti-PD-1 pharmacodynamics directly within tumors.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service