Skip to Content
Merck
All Photos(1)

Key Documents

SML2875

Sigma-Aldrich

DCC-2618

≥98% (HPLC)

Synonym(s):

DCC 2618, DCC2618, N-(2,5-Difluoro-4-(2-(1-methyl-1H-pyrazol-4-yl)pyridin-4-yloxy)phenyl)-N′-phenylcyclopropane-1,1-dicarboxamide, c-Kit-IN-1

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$118.00
25 MG
$475.00

$118.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$118.00
25 MG
$475.00

About This Item

Empirical Formula (Hill Notation):
C26H21F2N5O3
CAS Number:
Molecular Weight:
489.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$118.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(NC1=CC=CC=C1)C2(CC2)C(NC(C=C3F)=C(F)C=C3OC4=CC(C5=CN(C)N=C5)=NC=C4)=O

InChI

1S/C26H21F2N5O3/c1-33-15-16(14-30-33)21-11-18(7-10-29-21)36-23-13-19(27)22(12-20(23)28)32-25(35)26(8-9-26)24(34)31-17-5-3-2-4-6-17/h2-7,10-15H,8-9H2,1H3,(H,31,34)(H,32,35)

InChI key

WWOXKWLDMLMYQY-UHFFFAOYSA-N

Biochem/physiol Actions

DCC-2618 is an orally active, potent type II switch pocket (SP) inhibitor against c-KIT (IC50 = 4 nM/WT, 8 nM/V654A, 18 nM/T670I, 5 nM/D816H, 14 nM/D816V) and PDGFR. DCC-2618 locks c-KIT & PDGFR in an inactive conformation and is effective against multiple clinical forms of mutations resistant to type I ATP (catalytic)-site inhibitors Imatinib and Sunitinib. DCC-2618 inhibits cellular c-KIT activation (IC50 = 36 nM/WT, 2 nM/ex 11 del, 7 nM/ex 11 del & V654A, 53 nM/V560D & D820A) and is efficacious against KIT mutants-mediated cancer growth in cultres (IC50 = 2 nM; GIST with KIT D816Y) and in mice in vivo (50 mg/kg b.i.d. p.o.; GIST with KIT delW557K558/Y823D, AML with KIT N822K).
Orally active, potent type II switch pocket (SP) c-KIT & PDGFR inhibitor against multiple clinical forms of mutations resistant to Imatinib and Sunitinib.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathias Schneeweiss et al.
Haematologica, 103(5), 799-809 (2018-02-14)
Systemic mastocytosis is a complex disease defined by abnormal growth and accumulation of neoplastic mast cells in various organs. Most patients exhibit a D816V-mutated variant of KIT, which confers resistance against imatinib. Clinical problems in systemic mastocytosis arise from mediator-related
DCC-2618 is a potent inhibitor of wild-type and mutant KIT, including refractory Exon 17 D816 KIT mutations, and exhibits efficacy in refractory GIST and AML xenograft models, [abstract]
Smith BD, Hood MM, Wise SC, Kaufman MD, Lu WP, Rutkoski T, Flynn DL, Heinrich MC.
Cancer Research, 75 (15 Suppl), Abstract nr 2690-Abstract nr 2690 (2015)
Cancer discovery, 7(2), 121-122 (2017-01-13)
Tyrosine kinase inhibitors may initially control gastrointestinal stromal tumors, but most patients eventually experience disease progression due to activation loop mutations, which are resistant to approved drugs. However, phase I trials suggest that the cancer is sensitive to two new
DCC-2618, a pan KIT and PDGFR switch control inhibitor, achieves proof-of-concept in a first-in-human study
Janku F, George S, Razak A, Gordon M, Brooks D, Flynn DG, Kaufman M, Pitman J, Smith B, et al
European Journal of Cancer, 69, S4 (suppl-S4 (suppl (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service