Skip to Content
Merck
All Photos(1)

Key Documents

SML2841

Sigma-Aldrich

Tarenflurbil

≥98% (HPLC)

Synonym(s):

(R)-(−)-2-Fluoro-α-methyl-4-biphenylacetic acid, αR)-2-Fluoro-α-methyl[1,1′-biphenyl]-4-acetic acid, (-)-(R)-Flurbiprofen, (2R)-2-(3-Fluoro-4-phenylphenyl)propanoic acid, (R)-Flurbiprofen, Flurizan, R-flurbiprofen, (R)-2-Flurbiprofen, Tarenflurbil

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$37.10
50 MG
$147.00

$37.10


Estimated to ship onMay 20, 2025



Select a Size

Change View
10 MG
$37.10
50 MG
$147.00

About This Item

Linear Formula:
C6H5C6H3(F)CH(CH3)CO2H
CAS Number:
Molecular Weight:
244.26
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$37.10


Estimated to ship onMay 20, 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

C[C@@H](C(O)=O)c1ccc(c(F)c1)-c2ccccc2

InChI

1S/C15H13FO2/c1-10(15(17)18)12-7-8-13(14(16)9-12)11-5-3-2-4-6-11/h2-10H,1H3,(H,17,18)/t10-/m1/s1

InChI key

SYTBZMRGLBWNTM-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Tarenflurbil (or R-flurbiprofen) is an R enantiomer of racemic NSAID flurbiprofen that does not inhibit either cyclooxygenase 1 (COX-1) or cyclooxygenase 2 (COX-2). Tarenflurbil potently reduces levels of beta amyloid in human cells through direct inhibition of γ-secretase.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Repr. 2 - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason L Eriksen et al.
The Journal of clinical investigation, 112(3), 440-449 (2003-08-05)
Epidemiologic studies demonstrate that long-term use of NSAIDs is associated with a reduced risk for the development of Alzheimer disease (AD). In this study, 20 commonly used NSAIDs, dapsone, and enantiomers of flurbiprofen were analyzed for their ability to lower
T Morihara et al.
Journal of neurochemistry, 83(4), 1009-1012 (2002-11-08)
Non-steroidal anti-inflammatory drugs (NSAIDs) have been associated with reduced risk for Alzheimer's disease (AD) and selected NSAIDs racemates suppress beta-amyloid (Abeta) accumulation in vivo and Abeta42 production in vitro. Clinical use of NSAIDs for preventing or treating AD has been
Ling Rong Wong et al.
The Journal of pharmacy and pharmacology, 70(1), 59-69 (2017-10-17)
R-flurbiprofen (R-FP) was found to offer neuroprotective effects by inhibiting mitochondrial calcium overload induced by β-amyloid peptide toxicity in Alzheimer's disease (AD). However, poor brain penetration after oral administration posed a challenge to its further development for AD treatment. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service