Skip to Content
Merck
All Photos(1)

Key Documents

SML2823

Sigma-Aldrich

GT949

≥98% (HPLC)

Synonym(s):

3-[(4-Cyclohexyl-1-piperazinyl)[1-(2-phenylethyl)-1H-tetrazol-5-yl]methyl]-6-methoxy-2(1H)-quinolinone, GT 949, GT-949

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$117.00
25 MG
$327.00

$117.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$117.00
25 MG
$327.00

About This Item

Empirical Formula (Hill Notation):
C30H37N7O2
CAS Number:
Molecular Weight:
527.66
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$117.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C(C(C2=NN=NN2CCC3=CC=CC=C3)N4CCN(CC4)C5CCCCC5)=CC6=CC(OC)=CC=C6N1

Biochem/physiol Actions

GT949 is a potent and selective positive allosteric modulator (PAM) of the excitatory amino acid transporter EAAT2 (glutamate uptake EC50 = 0.26 nM; COS-7 EAAT2 transfectant), but not EAAT1 or EAAT3. GT949 enhances glutamate uptake of cultured rat astrocytes (EC50 = 1 nM, Emax = 158%) with no potency toward human serotonin (SERT), noradrenaline (NET) and dopamine (DAT) transporters, nor NMDA receptors (murine primary cortical neurons).
Potent and selective positive allosteric modulator (PAM) of the excitatory amino acid transporter EAAT2, but not EAAT1, EAAT3, NMDAR, SERT, NET, or DAT.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandhya Kortagere et al.
ACS chemical neuroscience, 9(3), 522-534 (2017-11-16)
Dysfunction of excitatory amino acid transporters (EAATs) has been implicated in the pathogenesis of various neurological disorders, such as stroke, brain trauma, epilepsy, and neurodegenerative diseases, among others. EAAT2 is the main subtype responsible for glutamate clearance in the brain
Romulo Martelli Falcucci et al.
ACS chemical neuroscience, 10(8), 3437-3453 (2019-07-02)
Dysfunction of excitatory amino acid transporters (EAATs) has been implicated in the pathogenesis of various neurological disorders, such as stroke, brain trauma, epilepsy, and several neurodegenerative disorders. EAAT2 is the main transporter subtype responsible for glutamate clearance in the brain
Rong-Wei Zhang et al.
Cell reports, 27(10), 2871-2880 (2019-06-06)
Retinal waves, the spontaneous patterned neural activities propagating among developing retinal ganglion cells (RGCs), instruct the activity-dependent refinement of visuotopic maps. Although it is known that the wave is initiated successively by amacrine cells and bipolar cells, the behavior and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service