Skip to Content
Merck
All Photos(1)

Documents

SML2808

Sigma-Aldrich

Sparsentan

≥98% (HPLC)

Synonym(s):

2-[4-[(2-Butyl-4-oxo-1,3-diazaspiro[4.4]non-1-en-3-yl)methyl]-2-(ethoxymethyl)phenyl]-N-(4,5-dimethyl-1,2-oxazol-3-yl)benzenesulfonamide, 4′-[(2-Butyl-4-oxo-1,3-diazaspiro[4.4]non-1-en-3-yl)methyl]-N-(4,5-dimethyl-3-isoxazolyl)-2′-(ethoxymethyl)[1,1′-biphenyl]-2-sulfonamide, BMS-346567, BMS346567, RE-021

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H40N4O5S
CAS Number:
Molecular Weight:
592.75
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Sparsentan (BMS346567) is an orally available and highly potent dual antagonist of angiotensin II AT1 receptor and endothelin A ETA receptor. It is an analog of BMS-248360 that is orally available in rats and higher species.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Howard Trachtman et al.
Journal of the American Society of Nephrology : JASN, 29(11), 2745-2754 (2018-10-27)
We evaluated and compared the effects of sparsentan, a dual endothelin type A (ETA) and angiotensin II type 1 receptor antagonist, with those of the angiotensin II type 1 receptor antagonist irbesartan in patients with primary FSGS. In this phase
Natesan Murugesan et al.
Journal of medicinal chemistry, 48(1), 171-179 (2005-01-07)
In a previous report we demonstrated that merging together key structural elements present in an AT(1) receptor antagonist (1, irbesartan) with key structural elements in a biphenylsulfonamide ET(A) receptor antagonist (2) followed by additional optimization provided compound 3 as a
Radko Komers et al.
Kidney international reports, 2(4), 654-664 (2017-11-17)
Primary focal segmental glomerulosclerosis (FSGS) is a leading cause of nephrotic syndrome and end-stage renal disease. There are no US Food and Drug Administration-approved therapies for FSGS, and treatment often fails to reduce proteinuria. Endothelin is an important factor in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service