Skip to Content
Merck
All Photos(1)

Key Documents

SML2784

Sigma-Aldrich

CMPD101 hydrochloride

≥98% (HPLC)

Synonym(s):

3-((4-Methyl-5-(pyridin-4-yl)-4H-1,2,4-triazol-3-yl)methylamino)-N-(2-(trifluoromethyl)benzyl)benzamide, 3-[[[4-Methyl-5-(4-pyridinyl)-4H-1,2,4-triazol-3-yl]methyl]amino]-N-[[2-(trifluoromethyl)phenyl]methyl]benzamide hydrochloride, Compound 101 hydrochloride, Cpd101 hydrochloride, Takeda101 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$162.00
25 MG
$652.00

$162.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$162.00
25 MG
$652.00

About This Item

Empirical Formula (Hill Notation):
C24H21N6OF3 · HCl
CAS Number:
Molecular Weight:
502.92
UNSPSC Code:
12352200
NACRES:
NA.77

$162.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN1C(CNC2=CC=CC(C(NCC3=CC=CC=C3C(F)(F)F)=O)=C2)=NN=C1C4=CC=NC=C4.Cl

InChI key

QBLGBXYDINNUFE-UHFFFAOYSA-N

Biochem/physiol Actions

Active site-targeting, potent and subtype-selective G protein-coupled receptor kinase GRK2/3 inhibitor with no inhibitory potency against GRK1/5.
CMPD101 is an active site-targeting, potent and subtype-selective G protein-coupled receptor kinase GRK2 & GRK3 inhibitor (human GRK2/3 IC50 = 54/32 nM with 3 μM ATP and tubulin dimer as substrate; bovine GRK2 IC50 = 290 nM with 0.5 mM ATP and bROS as substrate; no GRK1/5 inhibition at 125 μM). CMPD101 selectively inhibits GPR39 agonist-induced β-arrestin recruitment (by 94% at 10 μM against 30 μM GPR39-C3/50 μM ZnCl2), but not cAMP pathway desensitization in cultures and prevents β-arrestin2-biased D2R ligand UNC9994(1 μg/side bilateral local injection) from blocking NMDAR antagonist PCP (6 mg/kg i.p.)-induced locomotion (60%/12% blockage without/with 0.5 μg CMPD101 co-injection) in mice in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuji Shimizu et al.
Biochemical pharmacology, 140, 105-114 (2017-06-18)
GPR39, a G-protein-coupled receptor activated by zinc, reportedly activates multiple intracellular signaling pathways via Gs, Gq, G12/13, and β-arrestin, but little is known about downregulation of the receptor upon its activation. To our knowledge, this is the first report on
Seung-Ryoung Jung et al.
The Journal of general physiology, 147(3), 255-271 (2016-03-02)
Activated Gq protein-coupled receptors (GqPCRs) can be desensitized by phosphorylation and β-arrestin binding. The kinetics and individual contributions of these two mechanisms to receptor desensitization have not been fully distinguished. Here, we describe the shut off of protease-activated receptor 2
Arisbel B Gondin et al.
Frontiers in molecular neuroscience, 12, 104-104 (2019-05-24)
Differential regulation of the μ-opioid receptor (MOP) has been linked to the development of opioid tolerance and dependence which both limit the clinical use of opioid analgesics. At a cellular level, MOP regulation occurs via receptor phosphorylation, desensitization, plasma membrane
David M Thal et al.
Molecular pharmacology, 80(2), 294-303 (2011-05-21)
G protein-coupled receptors (GPCRs) are key regulators of cell physiology and control processes ranging from glucose homeostasis to contractility of the heart. A major mechanism for the desensitization of activated GPCRs is their phosphorylation by GPCR kinases (GRKs). Overexpression of
Alessandro Martini et al.
Neuropharmacology, 155, 76-88 (2019-05-28)
Neurotensin (NT) is a 13-amino acid peptide acting as a neuromodulator in the CNS. NT immunoreactive cell bodies, synaptic terminals and receptors (NTS) are intimately associated with the dopaminergic system. In fact, NT exerts a stimulatory action on the dopaminergic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service