Skip to Content
Merck
All Photos(1)

Key Documents

SML2779

Sigma-Aldrich

Tanaproget

≥98% (HPLC)

Synonym(s):

5-(1,4-Dihydro-4,4-dimethyl-2-thioxo-2H-3,1-benzoxazin-6-yl)-1-methyl-1H-pyrrole-2-carbonitrile, 5-(4,4-Dimethyl-2-thioxo-1,4-dihydro-2H-benzo[d][1,3]oxazin-6-yl)-1-methyl-1H-pyrrole-2-carbonitrile, NSP 989

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$193.00
25 MG
$772.00

$193.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$193.00
25 MG
$772.00

About This Item

Empirical Formula (Hill Notation):
C16H15N3OS
CAS Number:
Molecular Weight:
297.37
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$193.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

S=C1Nc2c(cc(cc2)c3[n](c(cc3)C#N)C)C(O1)(C)C

InChI

1S/C16H15N3OS/c1-16(2)12-8-10(4-6-13(12)18-15(21)20-16)14-7-5-11(9-17)19(14)3/h4-8H,1-3H3,(H,18,21)

InChI key

PYVFWTPEBMRKSR-UHFFFAOYSA-N

Biochem/physiol Actions

Non-steroidal progesterone receptor agonist
Tanaproget is a high affinity, high efficacy and selective orally available non-steroidal progesterone receptor agonist.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jody L Bapst et al.
Contraception, 74(5), 414-418 (2006-10-19)
This study aimed to evaluate the pharmacokinetics, pharmacodynamics and safety of the nonsteroidal progesterone receptor agonist, tanaproget. A randomized, double-blind, placebo-controlled, sequential-group study of ascending single doses of tanaproget was conducted in healthy, 25- to 45-year-old women on cycle days
Richard C Winneker et al.
Steroids, 73(7), 689-701 (2008-05-13)
Progesterone receptor (PR) modulators have evolved both structurally and mechanistically over the past half-century. Classical steroidal PR agonists continue to play an important role in women's health such as in oral contraception and post-menopausal hormone therapy whereas steroid-based PR antagonists
Louis Allott et al.
EJNMMI radiopharmacy and chemistry, 4(1), 1-1 (2019-10-30)
The histological evaluation of estrogen receptor (ER) and progesterone receptor (PR) expression in breast cancer lesions from biopsy tissue can stratify patients to receive endocrine therapy. Furthermore, PR expression can predict response to selective estrogen receptor modulators (SERMs). Current immunohistochemical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service