Skip to Content
Merck
All Photos(1)

Key Documents

SML2764

Sigma-Aldrich

Pitofenone hydrochloride

≥98% (HPLC)

Synonym(s):

2-[4-[2-(1-Piperidinyl)ethoxy]benzoyl]-benzoic acid methyl ester hydrochloride, Algifen Neo

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$83.40
25 MG
$337.00

$83.40

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$83.40
25 MG
$337.00

About This Item

Empirical Formula (Hill Notation):
C22H25NO4 · HCl
CAS Number:
Molecular Weight:
403.90
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$83.40

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[Cl-].N3(CCCCC3)CCOc1ccc(cc1)C(=O)c2c(cccc2)C(=O)OC.[H+]

InChI

1S/C22H25NO4.ClH/c1-26-22(25)20-8-4-3-7-19(20)21(24)17-9-11-18(12-10-17)27-16-15-23-13-5-2-6-14-23;/h3-4,7-12H,2,5-6,13-16H2,1H3;1H

InChI key

ZRFIFDFEDPJBII-UHFFFAOYSA-N

Biochem/physiol Actions

Pitofenone is a potent muscarinic receptor antagonist. Pitofenone in combination with diclofenac exhibits potent antispasmodic effect in cholinergic spasm.
potent muscarinic receptor antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K V Marthak et al.
Current medical research and opinion, 12(6), 366-373 (1991-01-01)
A single-blind, randomized clinical trial was carried out to compare the analgesic effectiveness in patients with renal colic of single intramuscular doses of diclofenac sodium (75 mg) versus a dipyrone (1 g)/spasmolytics combination, and diclofenac sodium (75 mg) versus pethidine
Shrinivas K Kulkarni et al.
Indian journal of experimental biology, 42(6), 567-569 (2004-07-21)
Biliary, ureteric and intestinal colic are extremely common clinical conditions associated with smooth muscle spasm. In the present study, antispasmodic activity was carried out against acetylcholine (10-640 ng/ml)-induced contractions on guinea pig ileum. Acetylcholine (10-640 ng/ml) induced concentration-dependent contraction of
J Sanahuja et al.
DICP : the annals of pharmacotherapy, 24(4), 361-364 (1990-04-01)
A comparative, randomized, double-blind study of diclofenac sodium 75 mg im versus Baralgin (a combination drug composed of dipyrone and two spasmolytics) 5 mL iv was performed on 57 patients with renal colic. Both groups were comparable as to age

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service