Skip to Content
Merck
All Photos(1)

Key Documents

SML2707

Sigma-Aldrich

TAS-102

≥98% (HPLC)

Synonym(s):

2′-Deoxy-5-trifluoromethyluridine for Trifluridine, Lonsurf, Tipiracil hydrochloride-trifluridine mixt, Trifluridine/Tipiracil HCl

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$196.00

$196.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$196.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

$196.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: 100 mg/mL
H2O: 15 mg/mL

storage temp.

2-8°C

General description

TAS-102 is a combination of two active ingredients: trifluridine (TFT), a nucleoside analog, and tipiracil (TPI), a thymidine phosphorylase inhibitor in a 1.0:0.5 molar ratio (TFT:TPI).1 TFT is the active antitumor component of TAS-102, its monophosphate form inhibits thymidylate synthase, an enzyme that plays a main role in DNA synthesis, and its triphosphate form is incorporated into DNA in tumor cells resulting in DNA dysfunction. TPI increases the bioavailability of TFT that is rapidly degraded to its inactive form by thymidine phosphorylase in the intestines and liver. The formulation presents a double targeted due to the cytotoxic effect of TFT, which is enhanced by TPI, while TPI also exerts antiangiogenic effects by inhibiting thymidine phosphorylase.2 TAS 102 inhibits KM20C colon cancer xenograft growth by 70.5% when was orally administered twice daily at a dose of 150 mg/kg for 14 days.3 TAS-102 has great potential to improve the treatment of malignancies when combined with other angiostatic or cytotoxic agents. A combination of TAS-102 and Avastin, an antineoplastic agent, was found to increase chemotherapeutic efficacy against colorectal cancer xenograft, compared with that for either drug alone.4

Biochem/physiol Actions

Anti cancer, Anti angiogenic

Other Notes

Solubility:DMSO 100 mg/mL; water 15mg/ml
TAS-102 is a mixture of 2 components. In order to maintain the original ratio between the components, the entire content of vial needs to be solubilized.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Muta. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service