Skip to Content
Merck
All Photos(1)

Key Documents

SML2684

Sigma-Aldrich

dTRIM24

≥98% (HPLC)

Synonym(s):

(2S,4R)-1-((S)-15-(tert-Butyl)-1-(3-(N-(1,3-dimethyl-2-oxo-6-(3-propoxyphenoxy)-2,3-dihydro-1H-benzo[d]imidazol-5-yl)sulfamoyl)phenyl)-1,13-dioxo-5,8,11-trioxa-2,14-diazahexadecan-16-oyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$117.00
5 MG
$445.00

$117.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$117.00
5 MG
$445.00

About This Item

Empirical Formula (Hill Notation):
C55H68N8O13S2
CAS Number:
Molecular Weight:
1113.30
UNSPSC Code:
12352200
NACRES:
NA.77

$117.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

ligand

VL-269

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(N1C)N(C)C2=C1C=C(OC3=CC=CC(OCCC)=C3)C(NS(C4=CC(C(NCCOCCOCCOCC(N[C@@H](C(C)(C)C)C(N5C[C@H](O)C[C@H]5C(NCC6=CC=C(C7=C(C)N=CS7)C=C6)=O)=O)=O)=O)=CC=C4)(=O)=O)=C2

InChI key

QUQTXFIMYFMULC-OGOZKGDESA-N

Biochem/physiol Actions

dTRIM24, a conjugate of IACS-9571 a TRIM24 bromodomain ligand and VL-269 a VHL E3 ubiquitin ligase ligand, is a selective bifunctional degrader of bromodomain-containing transcriptional regulator TRIM24 (transcriptional intermediary factor 1a). dTRIM24 induces rapid and sustain proteasome degradation of TRIM24 in 293FT cells.
selective bifunctional degrader of bromodomain-containing transcriptional regulator TRIM24

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lara N Gechijian et al.
Nature chemical biology, 14(4), 405-412 (2018-03-07)
The addressable pocket of a protein is often not functionally relevant in disease. This is true for the multidomain, bromodomain-containing transcriptional regulator TRIM24. TRIM24 has been posited as a dependency in numerous cancers, yet potent and selective ligands for the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service