Skip to Content
Merck
All Photos(1)

Key Documents

SML2674

Sigma-Aldrich

Ki20227

≥98% (HPLC)

Synonym(s):

Ki 20227, N-[4-[(6,7-Dimethoxy-4-quinolinyl)oxy]-2-methoxyphenyl]-N′-[1-(2-thiazolyl)ethyl]urea, N-{4-[(6,7-Dimethoxy-4-quinolinyl)oxy]-2-methoxyphenyl}-N′-[1-(1,3-thiazol-2-yl)ethyl]urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H24N4O5S
CAS Number:
Molecular Weight:
480.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c(ncc1)C(NC(=O)Nc2c(cc(cc2)Oc3c4c(ncc3)cc(c(c4)OC)OC)OC)C

InChI

1S/C24H24N4O5S/c1-14(23-26-9-10-34-23)27-24(29)28-17-6-5-15(11-20(17)30-2)33-19-7-8-25-18-13-22(32-4)21(31-3)12-16(18)19/h5-14H,1-4H3,(H2,27,28,29)

InChI key

SHPFDGWALWEPGS-UHFFFAOYSA-N

Biochem/physiol Actions

Ki20227 is an orally active, potent and selective M-CSF receptor c-Fms (CSF-1R) tyrosine kinase inhibitor (IC50 = 2 nM vs. 12 nM/KDR, 451 nM/c-Kit, 217 nM/PDGFβ; >1 μM/BTK, EGFR, FGFR2, FLT3, Fyn, Met, c-Src, PKA, PKCα) that inhibits M-CSF-dependent (50 ng/mL) c-Fms phosphorylation (by >90% at 10 nM; RAW264.7) and cell growth (IC50 = 14 nM; M-NFS-60). Ki20227 prevents osteolysis in a rat model of bone metastasis (50 mg/kg/day p.o.) by inhibiting A375 tumor-induced osteoclast formation and decreases the number of osteoclast-like cells on bone surfaces in ovariectomized rats (20 mg/kg/day p.o.) in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelda Chia et al.
eLife, 7 (2018-02-22)
It is now clear that microglia and macrophages are present in brain tumors, but whether or how they affect initiation and development of tumors is not known. Exploiting the advantages of the zebrafish (Danio rerio) model, we showed that macrophages
Igor Theurl et al.
Nature medicine, 22(8), 945-951 (2016-07-19)
Iron is an essential component of the erythrocyte protein hemoglobin and is crucial to oxygen transport in vertebrates. In the steady state, erythrocyte production is in equilibrium with erythrocyte removal. In various pathophysiological conditions, however, erythrocyte life span is compromised
Hiroaki Ohno et al.
European journal of immunology, 38(1), 283-291 (2007-12-19)
Macrophage colony-stimulating factor (M-CSF) is important in the development of macrophages and osteoclasts. Previous studies have also shown that CD11b(+) myeloblasts and osteoclasts play key roles during inflammation and bone destruction in arthritic lesions. In this study, we investigated whether
Hiroaki Ohno et al.
Molecular cancer therapeutics, 5(11), 2634-2643 (2006-11-24)
In bone metastatic lesions, osteoclasts play a key role in the development of osteolysis. Previous studies have shown that macrophage colony-stimulating factor (M-CSF) is important for the differentiation of osteoclasts. In this study, we investigated whether an inhibitor of M-CSF
Yasunori Uemura et al.
Journal of neuroimmunology, 195(1-2), 73-80 (2008-04-02)
Experimental autoimmune encephalomyelitis (EAE), an animal model of multiple sclerosis (MS), can be induced by the immunization of mice with myelin antigens in the form of myelin oligodendrocyte glycoprotein (MOG). Macrophage colony-stimulating factor (M-CSF) is required for the development of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service