Skip to Content
Merck
All Photos(1)

Key Documents

SML2651

Sigma-Aldrich

Elesclomol

≥98% (HPLC)

Synonym(s):

1,3-bis[2-Methyl-2-(phenylthioxomethyl)hydrazide]propanedioic acid, GSK842879A, N′1,N′3-Dimethyl-N′1,N′3-di(phenylcarbonothioyl)malonohydrazide, NSC 174939, NSC174939, STA 4783, STA-4783, STA4783

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$148.00
25 MG
$592.00

$148.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$148.00
25 MG
$592.00

About This Item

Empirical Formula (Hill Notation):
C19H20N4O2S2
CAS Number:
Molecular Weight:
400.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$148.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

S=C(N(NC(=O)CC(=O)NN(C)C(=S)c2ccccc2)C)c1ccccc1

InChI

1S/C19H20N4O2S2/c1-22(18(26)14-9-5-3-6-10-14)20-16(24)13-17(25)21-23(2)19(27)15-11-7-4-8-12-15/h3-12H,13H2,1-2H3,(H,20,24)(H,21,25)

InChI key

BKJIXTWSNXCKJH-UHFFFAOYSA-N

Biochem/physiol Actions

Copper (II)-chelating malonohydrazide derivative with in vitro and in vivo anticancer efficacy via reactive oxygen species (ROS) & apoptosis induction.
Elesclomol (STA-4783) is a copper (II)-chelating malonohydrazide derivative that exerts its in vitro and in vivo anticancer efficacy via reactive oxygen species (ROS)/oxidative stress & apoptosis induction in a Cu(II)-dependent manner (GI50 = 12 nM without vs. 129 nM with Cu2+ removal by TRIEN in K562 culture; typical conc. range 0.1-2 μM in cultures). Elesclomol chelates extracellular Cu(II) for mitochondria delivery, where upon Cu2+ reduction (to Cu+) & ROS generation, copper-free elesclomol then exits cells for continual Cu(II) shuttling and accumulation in mitochondria. In addition to Cu2+, elesclomol can also chelate Co2+ and Ni2+, but not Fe3+ or Zn2+.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masazumi Nagai et al.
Free radical biology & medicine, 52(10), 2142-2150 (2012-05-01)
Elesclomol is an investigational drug that exerts potent anticancer activity through the elevation of reactive oxygen species (ROS) levels and is currently under clinical evaluation as a novel anticancer therapeutic. Here we report the first description of selective mitochondrial ROS
Jia Yu Wang et al.
Cancer research, 77(22), 6226-6239 (2017-09-28)
MTH1 helps prevent misincorporation of ROS-damaged dNTPs into genomic DNA; however, there is little understanding of how MTH1 itself is regulated. Here, we report that MTH1 is regulated by polyubiquitination mediated by the E3 ligase Skp2. In melanoma cells, MTH1
Changyu Zhang et al.
Chemical communications (Cambridge, England), 54(31), 3835-3838 (2018-03-27)
A new fluorescence probe was developed for HClO detection under physiological conditions, which displayed fast response (t1/2 < 30 s) and large off-on fluorescence increase (>1000-fold) toward HClO with high sensitivity and selectivity. The probe was successfully applied for the
Yi-Jie Ren et al.
Frontiers in pharmacology, 8, 804-804 (2017-11-23)
In order to survive under conditions of low oxygen, cancer cells can undergo a metabolic switch to glycolysis and suppress mitochondrial respiration in order to reduce oxygen consumption and prevent excessive amounts of reactive oxygen species (ROS) production. Nucleus accumbens-1
Brian B Hasinoff et al.
Journal of inorganic biochemistry, 137, 22-30 (2014-05-07)
Elesclomol is an anticancer drug that is currently undergoing clinical trials. Elesclomol forms a strong 1:1 complex with Cu(II) and may exert its anticancer activity through the induction of oxidative stress and/or its ability to transport copper into the cell.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service