Skip to Content
Merck
All Photos(1)

Key Documents

SML2598

Sigma-Aldrich

Ataciguat

≥98% (HPLC)

Synonym(s):

5-Chloro-2-[[(5-chloro-2-thienyl)sulfonyl]amino]-N-[4-(4-morpholinylsulfonyl)phenyl]-benzamide, 5-Chloro-2-[[(5-chlorothien-2-yl)sulfonyl]amino]-N-[4-[(morpholin-4-yl)sulfonyl]phenyl]benzamide, HMR-1766, HMR1766

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$157.00
25 MG
$627.00

$157.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$157.00
25 MG
$627.00

About This Item

Empirical Formula (Hill Notation):
C21H19Cl2N3O6S3
CAS Number:
Molecular Weight:
576.49
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

$157.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[S](=O)(=O)(N4CCOCC4)c1ccc(cc1)NC(=O)c2c(ccc(c2)Cl)N[S](=O)(=O)c3[s]c(cc3)Cl

InChI

1S/C21H19Cl2N3O6S3/c22-14-1-6-18(25-34(28,29)20-8-7-19(23)33-20)17(13-14)21(27)24-15-2-4-16(5-3-15)35(30,31)26-9-11-32-12-10-26/h1-8,13,25H,9-12H2,(H,24,27)

InChI key

PQHLRGARXNPFCF-UHFFFAOYSA-N

Biochem/physiol Actions

Ataciguat (HMR-1766) is a soluble guanylate cyclase (sGC) activator
Soluble guanylate cyclase (sGC) activator

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ariane Migliato Martinelli et al.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques, 21(1), 38-45 (2018-02-17)
In endothelial cells, investigate if the soluble guanylate cyclase (sGC) activation or stimulation is able to potentiate the relaxation in vessels. Aortic and coronary rings with and without endothelium were placed in a myograph and cumulative concentration-effect curves for DETA-NO
Zongmin Zhou et al.
American journal of physiology. Heart and circulatory physiology, 295(4), H1763-H1771 (2008-09-02)
Many vascular diseases are characterized by increased levels of ROS that destroy the biological activity of nitric oxide and limit cGMP formation. In the present study, we investigated the cGMP-forming ability of HMR-1766 in cells exposed to oxidative stress. Pretreatment
Daniela Fraccarollo et al.
Basic research in cardiology, 109(4), 421-421 (2014-06-09)
Impaired nitric oxide (NO)-soluble guanylate cyclase (sGC)-cGMP signaling is involved in the pathogenesis of ischemic heart diseases, yet the impact of long-term sGC activation on progressive cardiac remodeling and heart failure after myocardial infarction (MI) has not been explored. Moreover

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service