Skip to Content
Merck
All Photos(1)

Key Documents

SML2548

Sigma-Aldrich

MS094

≥98% (HPLC)

Synonym(s):

2-[Methyl[[4-[4-(1-methylethoxy)phenyl]-1H-pyrrol-3-yl]methyl]amino]-ethanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$177.00
25 MG
$548.00

$177.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$177.00
25 MG
$548.00

About This Item

Empirical Formula (Hill Notation):
C17H24N2O2
CAS Number:
Molecular Weight:
288.38
UNSPSC Code:
51111800
NACRES:
NA.77

$177.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(CCO)CC1=CNC=C1C2=CC=C(OC(C)C)C=C2

InChI

1S/C17H24N2O2/c1-13(2)21-16-6-4-14(5-7-16)17-11-18-10-15(17)12-19(3)8-9-20/h4-7,10-11,13,18,20H,8-9,12H2,1-3H3

InChI key

GNAKYUUZAXUXKB-UHFFFAOYSA-N

Biochem/physiol Actions

Inactive control probe for PRMT inhibitor MS023
MS094 i an inactive control probe for PRMT inhibitor MS023. MS023 is a potent and selective chemical probe for Type I protein arginine methyltransferases (PRMTs) 1,3,4,6,and 8, which are responsible for asymmetric dimethylation of arginine residues. MS023 is active in cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduced C9orf72 expression exacerbates polyGR toxicity in patient iPSC-derived motor neurons and a Type I protein arginine methyltransferase inhibitor reduces that toxicity.
Dane, et al.
Frontiers in Cellular Neuroscience, 17, 1134090-1134090 (2023)
Mohammad S Eram et al.
ACS chemical biology, 11(3), 772-781 (2015-11-26)
Protein arginine methyltransferases (PRMTs) play a crucial role in a variety of biological processes. Overexpression of PRMTs has been implicated in various human diseases including cancer. Consequently, selective small-molecule inhibitors of PRMTs have been pursued by both academia and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service