Skip to Content
Merck
All Photos(1)

Key Documents

SML2543

Sigma-Aldrich

KA-11

≥98% (HPLC)

Synonym(s):

1-[1-Methyl-2-oxo-2-[4-[3-(trifluoromethyl)phenyl]-1-piperazinyl]ethyl]-2,5-pyrrolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20F3N3O3
CAS Number:
Molecular Weight:
383.36
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(N1C(CCC1=O)=O)C(N(CC2)CCN2C3=CC=CC(C(F)(F)F)=C3)=O

InChI

1S/C18H20F3N3O3/c1-12(24-15(25)5-6-16(24)26)17(27)23-9-7-22(8-10-23)14-4-2-3-13(11-14)18(19,20)21/h2-4,11-12H,5-10H2,1H3

InChI key

DWLZEEPJISTVEO-UHFFFAOYSA-N

Biochem/physiol Actions

Broad-spectrum anticonvulsant
KA-11 is a broad-spectrum anticonvulsant hybrid molecule that joins the chemical fragments of well-known anti-epileptic drugs such as ethosuximide, levetiracetam, and lacosamide. KA-11 was shown to have antiepileptogenic and antinociceptive properties in animal models. Its mechanism of action is believed to involve influence on the neuronal voltage-sensitive sodium and L-type calcium channels.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Socała et al.
ACS chemical neuroscience, 10(1), 636-648 (2018-09-25)
Recently, compound KA-11 was identified as a promising candidate for a new broad-spectrum anticonvulsant. This compound revealed wide protective activity across the most important animal models of seizures such as the maximal electroshock test (MES), the subcutaneous pentylenetetrazole test (
Krzysztof Kamiński et al.
Journal of medicinal chemistry, 58(13), 5274-5286 (2015-06-09)
The library of 27 new 1-(4-phenylpiperazin-1-yl)- or 1-(morpholin-4-yl)-(2,5-dioxopyrrolidin-1-yl)propanamides and (2,5-dioxopyrrolidin-1-yl)butanamides as potential new hybrid anticonvulsant agents was synthesized. These hybrid molecules join the chemical fragments of well-known antiepileptic drugs (AEDs) such as ethosuximide, levetiracetam, and lacosamide. Compounds 5, 10, 11

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service