Skip to Content
Merck
All Photos(1)

Key Documents

SML2526

Sigma-Aldrich

DDR-TRK-1

≥98% (HPLC)

Synonym(s):

(3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoro-methyl)-phenylamino)-(5-methyl-3-(pyrimidin-5-yl)-3-aza-bicyclo[4.4.0]deca-1(6),7,9-trien-9-yl)-methanone, (4R)-1,2,3,4-Tetrahydro-4-methyl-N-[3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-2-(5-pyrimidinyl)-7-isoquinolinecarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$193.00
25 MG
$509.00

$193.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$193.00
25 MG
$509.00

About This Item

Empirical Formula (Hill Notation):
C26H23F3N6O
CAS Number:
Molecular Weight:
492.50
UNSPSC Code:
12352200
NACRES:
NA.77

$193.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -25 to -20°, c = 0.5 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

A chemical probe for the DDR and TRK kinase
DDR-TRK-1 was shown to inhibit the kinase activity of Discoidin Domain Receptor 1 (DDR1) with an IC50 value of 9.4 nM. DDR receptors are linked to the progression of various human diseases, including fibrotic disorders such as pulmonary fibrosis, as well as atherosclerosis and cancer. DDR-TRK-1 showed a therapeutic effect in a mouse model of pulmonary fibrosis and inhibited Panc-1 pancreatic cancer cell colony formation and migration. DDR-TRK-1 also inhibits the TRK family kinases TRKA, TRKB, and TRKC, which are selectively expressed in neuronal tissue. DDR-TRK-1N is the negative control compound.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen Wang et al.
Journal of medicinal chemistry, 59(12), 5911-5916 (2016-05-25)
The structure-based design of 1, 2, 3, 4-tetrahydroisoquinoline derivatives as selective DDR1 inhibitors is reported. One of the representative compounds, 6j, binds to DDR1 with a Kd value of 4.7 nM and suppresses its kinase activity with an IC50 value

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service