Skip to Content
Merck
All Photos(1)

Key Documents

SML2509

Sigma-Aldrich

Fenticonazole Nitrate

Synonym(s):

Fenticonazole nitrate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$207.00

$207.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$207.00

About This Item

Empirical Formula (Hill Notation):
C24H21Cl2N3O4S
CAS Number:
Molecular Weight:
518.41
UNSPSC Code:
12352200
NACRES:
NA.77

$207.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

form

powder

storage temp.

2-8°C

SMILES string

S(c2ccc(cc2)COC(C[n]4cncc4)c3c(cc(cc3)Cl)Cl)c1ccccc1.[N+](=O)([O-])O

InChI

1S/C24H20Cl2N2OS.HNO3/c25-19-8-11-22(23(26)14-19)24(15-28-13-12-27-17-28)29-16-18-6-9-21(10-7-18)30-20-4-2-1-3-5-20;2-1(3)4/h1-14,17,24H,15-16H2;(H,2,3,4)

InChI key

FJNRUWDGCVDXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Fenticonazole is an imidazole antifungal drug.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service