Skip to Content
Merck
All Photos(1)

Key Documents

SML2461

Sigma-Aldrich

LM22B-10

≥98% (HPLC)

Synonym(s):

2,2′,2′′,2′′′-[[(4-Chlorophenyl)methylene]bis(4,1-phenylenenitrilo)]tetrakis[ethanol], 2,2′,2′,2′′′-[[(4-Chlorophenyl)methylene]bis(4,1-phenylenenitrilo)]tetrakisethanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$109.00
25 MG
$439.00

$109.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$109.00
25 MG
$439.00

About This Item

Empirical Formula (Hill Notation):
C27H33ClN2O4
CAS Number:
Molecular Weight:
485.01
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$109.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

faint blue to green

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC1=CC=C(C(C2=CC=C(N(CCO)CCO)C=C2)C3=CC=C(N(CCO)CCO)C=C3)C=C1

InChI key

QCXQLSGBOUUVNH-UHFFFAOYSA-N

Biochem/physiol Actions

LM22B-10 is a brain penetrant, potent and selective TrkB and TrkC neurotrophin receptors agonist that improves cell survival and potently accelerated neurite outgrowth. LM22B-10 inhibits binding of BDNF to TrkB-expressing cells and NT-3 to TrkC-expressing cells.
brain penetrant, potent and selective TrkB and TrkC agonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Yang et al.
Neuropharmacology, 110(Pt A), 343-361 (2016-06-24)
Neurotrophin (NT) receptors are coupled to numerous signaling networks that play critical roles in neuronal survival and plasticity. Several non-peptide small molecule ligands have recently been reported that bind to and activate specific tropomyosin-receptor kinase (Trk) NT receptors, stimulate their
Tao Zhang et al.
Asian Pacific journal of tropical medicine, 10(12), 1172-1176 (2017-12-23)
To study the influence of glycyrrhetinic acid (GA) on bronchial asthma (BA) smooth muscle proliferation and apoptosis as well as inflammatory factor expression and its molecular mechanism. Male SD guinea pigs were selected and made into asthma models, bronchial asthma

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service