Skip to Content
Merck
All Photos(1)

Key Documents

SML2457

Sigma-Aldrich

Amgen16

≥98% (HPLC)

Synonym(s):

1-((1-(2-Amino-5-chloropyrimidin-4-yl)indolin-6-yl)ethynyl)cyclopentanol, 1-[2-[1-(2-Amino-5-chloro-4-pyrimidinyl)-2,3-dihydro-1H-indol-6-yl]ethynyl]cyclopentanol, Amgen 16

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$123.00
25 MG
$496.00

$123.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$123.00
25 MG
$496.00

About This Item

Empirical Formula (Hill Notation):
C19H19ClN4O
CAS Number:
Molecular Weight:
354.83
UNSPSC Code:
12352200
NACRES:
NA.77

$123.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=NC=C(C(N2CCC3=C2C=C(C=C3)C#CC4(CCCC4)O)=N1)Cl

InChI

1S/C19H19ClN4O/c20-15-12-22-18(21)23-17(15)24-10-6-14-4-3-13(11-16(14)24)5-9-19(25)7-1-2-8-19/h3-4,11-12,25H,1-2,6-8,10H2,(H2,21,22,23)

InChI key

KJIQYGRWNLGGMI-UHFFFAOYSA-N

Biochem/physiol Actions

Amgen16 is an NF-κB-inducing kinase (NIK) inhibitor that selectively disrupts noncanonical NF-κB activation in U-2 OS cells (lymphotoxin β receptor LTβR-/TWEAK recetor FN14-mediated p52 nuclear translocation IC50 = 417 nM/2.517 μM) without affecting canonical NF-κB activation (TNF-α-stimulated p65 (RelA) nuclear translocation IC50 >30 μM). Likewise, Amgen16 (1 μM, 30 min pretreatment) selectively inhibits NFKB2 (p100) mRNA production upon noncanonical NF-κB activation (by 84%; 20 ng/mL TWEAK for 4 h), while exhibiting much smaller effect against canonical NF-κB stimulator-induced NFKB2 production (by 36%; 10 ng/mLTNFα for 4 h).
NF-κB-inducing kinase (NIK) inhibitor that selectively disrupts noncanonical over canonical NF-κB activation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kate L Henry et al.
Science signaling, 11(541) (2018-08-02)
Members of the family of nuclear factor κB (NF-κB) transcription factors are critical for multiple cellular processes, including regulating innate and adaptive immune responses, cell proliferation, and cell survival. Canonical NF-κB complexes are retained in the cytoplasm by the inhibitory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service